Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • MMP14 Antibodies

          Proteintech

          MT1-MMP Polyclonal Antibody

          28 References
          View all (18) MMP14 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MT1-MMP Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          MT1-MMP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MT1-MMP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MT1-MMP Antibody (14552-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using (Product # 14552-1-AP) (MT1-MMP antibody) at dilution of 1:50 and Alexa Fluor 488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MT1-MMP Antibody in Immunocytochemistry (ICC/IF)
          MT1-MMP Antibody in Immunocytochemistry (ICC/IF)
          MT1-MMP Antibody in Western Blot (WB)
          MT1-MMP Antibody in Western Blot (WB)
          MT1-MMP Antibody in Western Blot (WB)
          MT1-MMP Antibody in Western Blot (WB)
          MT1-MMP Polyclonal Antibody

          Product Details

          14552-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 20 publications 20 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MT1-MMP Fusion Protein Ag5965 (1-350 aa encoded by BC064803)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          30 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSPAPRPSR CLLLPLLTLG TALASLGSAQ SSSFSPEAWL QQYGYLPPGD LRTHTQRSPQ SLSAAIAAMQ KFYGLQVTGK ADADTMKAMR RPRCGVPDKF GAEIKANVRR KRYAIQGLKW QHNEITFCIQ NYTPKVGEYA TYEAIRKAFR VWESATPLRF REVPYAYIRE GHEKQADIMI FFAEGFHGDS TPFDGEGGFL AHAYFPGPNI GGDTHFDSAE PWTVRNEDLN GNDIFLVAVH ELGHALGLEH SSDPSAIMAP FYQWMDTENF VLPDDDRRGI QQLYGGESGF PTKMPPQPRT TSRPSVPDKP KNPTYGPNIC DGNFDTVAML RGEMFVFKER WFWRVRNNQV M (1-350 aa encoded by BC064803)

          Target Information

          Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; Matrix metalloproteinase-14; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; Membrane-type matrix metalloproteinase 1; Membrane-type-1 matrix metalloproteinase; MMP; MMP-14; MMP-X MT-MMP; MMP-X1; MMPs; MT-MMP; MT-MMP 1; MT-MMP MT1-MMP; MT1-MMP; MT1MMP; MTMMP1; type 1 matrix metalloprotease 14; unnamed protein product

          View more View less

          Gene Aliases: AI325305; MMP-14; MMP-X1; MMP14; MT-MMP; MT-MMP 1; MT-MMP-1; MT1-MMP; MT1MMP; Mtmmp; MTMMP1; sabe; WNCHRS

          View more View less

          UniProt ID: (Human) P50281, (Mouse) P53690, (Rat) Q10739

          View more View less

          Entrez Gene ID: (Human) 4323, (Mouse) 17387, (Rat) 81707

          View more View less

          Function(s)
          endopeptidase activity metalloendopeptidase activity serine-type endopeptidase activity integrin binding protein binding peptidase activity metallopeptidase activity zinc ion binding hydrolase activity metal ion binding metalloaminopeptidase activity transcription factor activity, sequence-specific DNA binding calcium ion binding peptidase activator activity metalloprotease
          Process(es)
          skeletal system development ossification angiogenesis ovarian follicle development response to hypoxia endothelial cell proliferation endochondral ossification proteolysis response to oxidative stress male gonad development response to mechanical stimulus anatomical structure morphogenesis response to hormone positive regulation of myotube differentiation positive regulation of protein processing cell migration protein processing extracellular matrix disassembly protein catabolic process extracellular matrix organization positive regulation of cell growth lung development positive regulation of cell migration collagen catabolic process zymogen activation endodermal cell differentiation chondrocyte proliferation astrocyte cell migration response to estrogen positive regulation of B cell differentiation negative regulation of Notch signaling pathway embryonic cranial skeleton morphogenesis branching morphogenesis of an epithelial tube tissue remodeling cell motility negative regulation of focal adhesion assembly head development bone development cellular response to genistein craniofacial suture morphogenesis GDF15-GFRAL signaling pathway negative regulation of GDF15-GFRAL signaling pathway regulation of protein localization to plasma membrane positive regulation of macrophage migration response to odorant regulation of transcription, DNA-templated positive regulation of peptidase activity response to organic cyclic compound
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline