Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • MTMR4 Antibodies

          Proteintech

          MTMR4 Polyclonal Antibody

          1 Reference
          View all (4) MTMR4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MTMR4 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          MTMR4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MTMR4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MTMR4 Antibody (13488-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells using 13488-1-AP (MTMR4 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MTMR4 Antibody in Immunocytochemistry (ICC/IF)
          MTMR4 Antibody in Western Blot (WB)
          MTMR4 Antibody in Western Blot (WB)
          MTMR4 Antibody in Western Blot (WB)
          MTMR4 Polyclonal Antibody

          Product Details

          13488-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MTMR4 Fusion Protein Ag4310 (845-1195 aa encoded by BC035609)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.15 mg/mL

          Amount

          22.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: SHQEQLSSV PDLTHGEEDI GKRGNNRNGQ LLENPRFGKM PLELVRKPIS QSQISEFSFL GSNWDSFQGM VTSFPSGEAT PRRLLSYGCC SKRPNSKQMR ATGPCFGGQW AQREGVKSPV CSSHSNGHCT GPGGKNQMWL SSHPKQVSST KPVPLNCPSP VPPLYLDDDG LPFPTDVIQH RLRQIEAGYK QEVEQLRRQV RELQMRLDIR HCCAPPAEPP MDYEDDFTCL KESDGSDTED FGSDHSEDCL SEASWEPVDK KETEVTRWVP DHMASHCYNC DCEFWLAKRR HHCRNCGNVF CAGCCHLKLP IPDQQLYDPV LVCNSCYEHI QVSRARELMS QQLKKPIATA SS (845-1195 aa encoded by BC035609)

          Target Information

          MTMR, a member of the myotubularin family, encodes an intracellular protein of 1, 195 amino acids, and shares approximately 47% sequence identity with MTMR3, spiking to over 70% identity in the catalytic domain and the double zinc finger (FYVE) motif. PCR analysis detects expression of MTMR4 in all human tissues examined except lung, small intestine, stomach, salivary gland, adrenal gland, and uterus. Subcellular localization of MTMR4 in transfected HeLa cells showed a perinuclear distribution, similar to that seen with MTMR3. MTMR4 dephosphorylates para-nitrophenylphosphate and phosphatidylinositol 3-phosphate.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: DKFZp781E194; FYVE domain-containing dual specificity protein phosphatase 2; FYVE zinc finger phosphatase; FYVE-DSP2; MGC163449; MGC163451; MTMR7; Myotubularin-related protein 4; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase MTMR4; Phosphatidylinositol-3-phosphate phosphatase; Zinc finger FYVE domain-containing protein 11; zinc finger, FYVE domain containing 11

          View more View less

          Gene Aliases: AA596759; ESTM44; FYVE-DSP2; KIAA0647; mKIAA0647; MTMR4; ZFYVE11

          View more View less

          UniProt ID: (Human) Q9NYA4, (Mouse) Q91XS1

          View more View less

          Entrez Gene ID: (Human) 9110, (Mouse) 170749

          View more View less

          Function(s)
          phosphatidylinositol-3-phosphatase activity protein serine/threonine phosphatase activity protein tyrosine phosphatase activity protein binding zinc ion binding hydrolase activity protein phosphatase binding metal ion binding phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity binding, bridging R-SMAD binding phosphoprotein phosphatase activity phosphatase activity phosphatase
          Process(es)
          phosphatidylinositol biosynthetic process negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of BMP signaling pathway phosphatidylinositol dephosphorylation midbody abscission phagosome maturation negative regulation of endocytic recycling dephosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline