Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MVP Antibodies

          Proteintech

          MVP/LRP Polyclonal Antibody

          12 References
          View all (37) MVP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MVP/LRP Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (21)
          MVP/LRP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MVP/LRP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MVP/LRP Antibody (16478-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using MVP antibody 16478-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MVP/LRP Antibody in Immunocytochemistry (ICC/IF)
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Western Blot (WB)
          MVP/LRP Antibody in Immunoprecipitation (IP)
          MVP/LRP Polyclonal Antibody

          Product Details

          16478-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MVP/LRP Fusion Protein Ag9593 (544-893 aa encoded by BC015623)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: NWHFEVNDR KDPQETAKLF SVPDFVGDAC KAIASRVRGA VASVTFDDFH KNSARIIRTA VFGFETSEAK GPDGMALPRP RDQAVFPQNG LVVSSVDVQS VEPVDQRTRD ALQRSVQLAI EITTNSQEAA AKHEAQRLEQ EARGRLERQK ILDQSEAEKA RKELLELEAL SMAVESTGTA KAEAESRAEA ARIEGEGSVL QAKLKAQALA IETEAELQRV QKVRELELVY ARAQLELEVS KAQQLAEVEV KKFKQMTEAI GPSTIRDLAV AGPEMQVKLL QSLGLKSTLI TDGSTPINLF NTAFGLLGMG PEGQPLGRRV ASGPSPGEGI SPQSAQAPQA PGDNHVVPVL R (544-893 aa encoded by BC015623)

          Target Information

          This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: lrp; Lung resistance-related protein; Major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; unnamed protein product; VAULT1; vRNP

          View more View less

          Gene Aliases: 2310009M24Rik; LRP; MVP; VAULT1

          View more View less

          UniProt ID: (Human) Q14764, (Rat) Q62667, (Mouse) Q9EQK5

          View more View less

          Entrez Gene ID: (Human) 9961, (Rat) 64681, (Mouse) 78388

          View more View less

          Function(s)
          protein binding protein kinase binding protein phosphatase binding identical protein binding scaffold/adaptor protein
          Process(es)
          cell proliferation protein transport negative regulation of protein autophosphorylation ERBB signaling pathway negative regulation of epidermal growth factor receptor signaling pathway mRNA transport negative regulation of protein tyrosine kinase activity protein activation cascade negative regulation of signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline