Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: CELCGNSYTD IKNLKKHKTK VHSGADKTLD SSAEDHTLSE QDSIQKSPLS ETMDVKPSDM TLPLALPLGT EDHHMLLPVT DTQSPTSDTL LRSTVNGYSE PQLIFLQQLY
This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myoneurin; Zinc finger and BTB domain-containing protein 31; zinc finger protein with BTB/POZ domain
Gene Aliases: MYNN; OSZF; SBBIZ1; ZBTB31; ZNF902
UniProt ID: (Human) Q9NPC7
Entrez Gene ID: (Human) 55892
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support