Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ALFTYESLK KTKCREFVPS RDEIEALIHR QEMTSTVYCH GGGSCKITIN SHTTAGEVVE KLIRGLAMED SRNMFALFEY NGHVDKAIES RTVVADVLAK FEKLAATSEV GDLPWKFYFK LYCFLDTDNV PKDSVEFAFM FEQAHEAVIH GHHPAPEENL QVLAALRLQY LQGDYTLHAA IPPLEEVYSL QRLKARISQS TKTFTPCERL EKRRTSFLEG TLRRSFRTGS VVRQKVEEEQ MLDMWIKEEV (1669-1917 aa encoded by BC137168)
This gene encodes a member of the myosin superfamily. The protein represents an unconventional myosin; it should not be confused with the conventional non-muscle myosin-10 (MYH10). Unconventional myosins contain the basic domains of conventional myosins and are further distinguished from class members by their tail domains. This gene functions as an actin-based molecular motor and plays a role in integration of F-actin and microtubule cytoskeletons during meiosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myo X; unconventional myosin; unconventional myosin; unconventional myosin-10; unconventionnal myosin-X; unnamed protein product
Gene Aliases: D15Ertd600e; mKIAA0799; myosin-X; MyoX
UniProt ID: (Human) Q9HD67, (Mouse) F8VQB6
Entrez Gene ID: (Human) 4651, (Mouse) 17909
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support