Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LHMACASGY KEVVSLILEH GGDLNIVDDQ YWTPLHLAAK YGQTNLVKLL LMHQANPHLV NCNEEKASDI AASEFIEEML LKAEIAWEEK MKEPLSASTL AQEEPYEEII HDLPVLSSKL SPLVLPIAKQ DSLLEKDIMF KDATKGLCKQ QSQDSIPENP MMSGSTKPEQ VKLMPPAPND DLATLS (226-410 aa encoded by BC146791)
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments. May be involved in targeting of the catalytic subunit of protein phosphatase 1 during brain development. Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MYO16 variant protein; myo16 {ECO:0000250|UniProtKB:Q9Y6X6}; Myosin heavy chain myr 8; myosin-XVI; Neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 3; protein phosphatase 1, regulatory subunit 107; Unconventional myosin-16; Unconventional myosin-XVI; unnamed protein product
Gene Aliases: KIAA0865; MYAP3; MYO16; MYO16B; MYR8; NYAP3; PPP1R107
UniProt ID: (Human) Q9Y6X6, (Rat) Q9ERC1
Entrez Gene ID: (Human) 23026, (Rat) 192253
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support