Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MYOD Antibodies

          Proteintech

          MYOD1 Polyclonal Antibody

          76 References
          View all (47) MYOD antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MYOD1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (22)
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MYOD1 Antibody (18943-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded mouse embryo tissue slide using 18943-1-AP ( MYOD1 Antibody) at dilution of 1:200 (under 40x lens). heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MYOD1 Antibody in Western Blot (WB)
          MYOD1 Antibody in Western Blot (WB)
          MYOD1 Antibody in Western Blot (WB)
          MYOD1 Antibody in Western Blot (WB)
          MYOD1 Antibody in Western Blot (WB)
          MYOD1 Antibody in Western Blot (WB)
          MYOD1 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MYOD1 Polyclonal Antibody

          Product Details

          18943-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 51 publications 51 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 14 publications 14 publications

          Flow Cytometry (Flow)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MYOD1 Fusion Protein Ag13512 (1-320 aa encoded by BC064493)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          30 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MELLSPPLR DVDLTAPDGS LCSFATTDDF YDDPCFDSPD LRFFEDLDPR LMHVGALLKP EEHSHFPAAV HPAPGAREDE HVRAPSGHHQ AGRCLLWACK ACKRKTTNAD RRKAATMRER RRLSKVNEAF ETLKRCTSSN PNQRLPKVEI LRNAIRYIEG LQALLRDQDA APPGAAAAFY APGPLPPGRG GEHYSGDSDA SSPRSNCSDG MMDYSGPPSG ARRRNCYEGA YYNEAPSEPR PGKSAAVSSL DCLSSIVERI STESPAAPAL LLADVPSESP PRRQEAAAPS EGESSGDPTQ SPDAAPQCPA GANPNPIYQV L (1-320 aa encoded by BC064493)

          Target Information

          The Myogenic determination gene (MyoD) was first identified by the virtue of its ability to convert embryonic mouse fibroblast cells to muscle cells. It was subsequently shown that forced expression of MyoD (human homolog is myf 3) gene in a wide variety of normal and neoplastic cells could either convert the cells to muscle cells or activate a set of the otherwise transcriptionally inactive muscle-specific genes in these cells. The regulatory domain of the MyoD gene product lies within a 70 amino acid region and comprises a basic DNA binding motif and a helix-loop-helix (HLH) dimerization motif. Subsequent studies identified three other genes whose products shared sequence homology for the basic HLH domain of MyoD. These are; myf5, myogenin (human homolog is myf4) and myf6 (also known as MRF4 and herculin).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: class C basic helix-loop-helix protein 1; myf-3; MyoD; MyoD1; myogenic factor 3; unnamed protein product

          View more View less

          Gene Aliases: bHLHc1; CMYO17; CMYP17; MYF3; MYOD; Myod-1; MYODRIF; PUM

          View more View less

          UniProt ID: (Human) P15172, (Rat) Q02346, (Mouse) P10085

          View more View less

          Entrez Gene ID: (Human) 4654, (Rat) 337868, (Mouse) 17927

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding ligand-dependent nuclear receptor binding enzyme binding chromatin DNA binding ubiquitin protein ligase binding protein homodimerization activity bHLH transcription factor binding sequence-specific DNA binding protein dimerization activity RNA polymerase II sequence-specific DNA binding transcription factor binding E-box binding sequence-specific double-stranded DNA binding promoter-specific chromatin binding
          Process(es)
          regulation of alternative mRNA splicing, via spliceosome regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter muscle organ development myoblast fate determination skeletal muscle tissue development myoblast fusion cellular response to starvation regulation of gene expression skeletal muscle atrophy myotube differentiation myotube cell development myotube differentiation involved in skeletal muscle regeneration skeletal muscle cell differentiation response to muscle stretch muscle cell differentiation protein unfolding skeletal muscle tissue regeneration positive regulation of skeletal muscle tissue regeneration regulation of RNA splicing skeletal muscle fiber adaptation myoblast differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter skeletal muscle fiber development positive regulation of skeletal muscle fiber development striated muscle cell differentiation positive regulation of muscle cell differentiation cellular response to tumor necrosis factor cellular response to glucocorticoid stimulus cellular response to estradiol stimulus cellular response to oxygen levels positive regulation of myoblast fusion positive regulation of snRNA transcription by RNA polymerase II negative regulation of myoblast proliferation tissue development muscle cell fate commitment animal organ development cardiac muscle cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-gw77j:80/100.66.76.64:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0