Product References
Signal-dependent slow leukocyte rolling does not require cytoskeletal anchorage of P-selectin glycoprotein ligand-1 (PSGL-1) or integrin αLβ2.
The Journal of biological chemistry
Shao B,Yago T,Coghill PA,Klopocki AG,Mehta-D'souza P,Schmidtke DW,Rodgers W,McEver RP
MA5-14043 was used in western blot to study whether cytoskelatal-anchorage of PSGL-1 and alphaLbeta2 intergrin is required for signal-dependent slow leukocyte rolling
Fri Jun 01 00:00:00 EDT 2012
Signal-dependent slow leukocyte rolling does not require cytoskeletal anchorage of P-selectin glycoprotein ligand-1 (PSGL-1) or integrin αLβ2.
The Journal of biological chemistry
Shao B,Yago T,Coghill PA,Klopocki AG,Mehta-D'souza P,Schmidtke DW,Rodgers W,McEver RP
MA5-14043 was used in western blot to study whether cytoskelatal-anchorage of PSGL-1 and alphaLbeta2 intergrin is required for signal-dependent slow leukocyte rolling
Fri Jun 01 00:00:00 EDT 2012
Cytoplasmic ezrin and moesin correlate with poor survival in head and neck squamous cell carcinoma.
Head and neck pathology
Schlecht NF,Brandwein-Gensler M,Smith RV,Kawachi N,Broughel D,Lin J,Keller CE,Reynolds PA,Gunn-Moore FJ,Harris T,Childs G,Belbin TJ,Prystowsky MB
MA5-14043 was used in immunohistochemistry to study the prognostic value of cytoplasmic expression of ezrin and moesin in head and neck squamous cell carcinoma
Fri Jun 01 00:00:00 EDT 2012
Distinct cellular and therapeutic effects of obatoclax in rituximab-sensitive and -resistant lymphomas.
British journal of haematology
Brem EA,Thudium K,Khubchandani S,Tsai PC,Olejniczak SH,Bhat S,Riaz W,Gu J,Iqbal A,Campagna R,Knight J,Mavis C,Hoskin P,Deeb G,Gibbs JF,Fetterly G,Czuczman MS,Hernandez-Ilizaliturri FJ
MA5-14043 was used in western blot to study the cellular and therapeutic effects of obatoclax in rituximab-sensitive and -resistant lymphomas
Wed Jun 01 00:00:00 EDT 2011
The effect of sphingomyelin synthase 2 (SMS2) deficiency on the expression of drug transporters in mouse brain.
Biochemical pharmacology
Zhang Y,Dong J,Zhu X,Wang W,Yang Q
MA5-14043 was used in western blot to identify the relationship between sphingomyelin synthetase 2 levels and drug transporter expression
Mon Aug 01 00:00:00 EDT 2011
Distinct molecular mechanisms responsible for bortezomib-induced death of therapy-resistant versus -sensitive B-NHL cells.
Blood
Olejniczak SH,Blickwedehl J,Belicha-Villanueva A,Bangia N,Riaz W,Mavis C,Clements JL,Gibbs J,Hernandez-Ilizaliturri FJ,Czuczman MS
MA5-14043 was used in western blot to study the molecular mechanisms responsible for bortezomib-induced death of therapy-resistant versus -sensitive B-NHL cells
Thu Dec 16 00:00:00 EST 2010
Immunohistochemical profiling of node negative breast carcinomas allows prediction of metastatic risk.
International journal of oncology
Giusiano S,Secq V,Carcopino X,Carpentier S,Andrac L,Lavaut MN,Allasia C,Bonnier P,Iovanna JL,Garcia S,Boubli L,Birnbaum D,Charpin C
MA5-14043 was used in immunohistochemistry - paraffin section to identify a prognostic immunohistochemical signature indicative of risk of early metastasis in node-negative breast carcinomas and use this information to develop targeted therapies
Thu Apr 01 00:00:00 EDT 2010
TREM-like transcript-1 protects against inflammation-associated hemorrhage by facilitating platelet aggregation in mice and humans.
The Journal of clinical investigation
Washington AV,Gibot S,Acevedo I,Gattis J,Quigley L,Feltz R,De La Mota A,Schubert RL,Gomez-Rodriguez J,Cheng J,Dutra A,Pak E,Chertov O,Rivera L,Morales J,Lubkowski J,Hunter R,Schwartzberg PL,McVicar DW
MA5-14043 was used in western blot to investigate the protective effect of TREM-like transcript-1 -1 against inflammation-associated hemorrhage
Mon Jun 01 00:00:00 EDT 2009
Anti-moesin antibodies in the serum of patients with aplastic anemia stimulate peripheral blood mononuclear cells to secrete TNF-alpha and IFN-gamma.
Journal of immunology (Baltimore, Md. : 1950)
Takamatsu H,Espinoza JL,Lu X,Qi Z,Okawa K,Nakao S
MA5-14043 was used in flow cytometry to study the role of anti-moesin antibodies in the secretion of TNF-alpha and IFN-gamma by PBMCs in patients with aplastic anemia
Thu Jan 01 00:00:00 EST 2009
Acquired resistance to rituximab is associated with chemotherapy resistance resulting from decreased Bax and Bak expression.
Clinical cancer research : an official journal of the American Association for Cancer Research
Olejniczak SH,Hernandez-Ilizaliturri FJ,Clements JL,Czuczman MS
MA5-14043 was used in western blot to study the role of Bax and Bak downregulation in the drug resistance to rituximab
Sat Mar 01 00:00:00 EST 2008
Specific antibodies to moesin, a membrane-cytoskeleton linker protein, are frequently detected in patients with acquired aplastic anemia.
Blood
Takamatsu H,Feng X,Chuhjo T,Lu X,Sugimori C,Okawa K,Yamamoto M,Iseki S,Nakao S
MA5-14043 was used in western blot to identify novel auto-antibodies in acquired aplastic anemia
Thu Mar 15 00:00:00 EDT 2007
Adhesion molecules as prognostic markers in pancreatic adenocarcinoma.
Journal of surgical oncology
Torer N,Kayaselcuk F,Nursal TZ,Yildirim S,Tarim A,Nòyan T,Karakayali H
MA5-14043 was used in immunohistochemistry to study the prognostic value of ezrin, radixin, moesin and E-cadherin in pancreatic ductal adenocarcinoma
Mon Oct 01 00:00:00 EDT 2007
Differential tissue and subcellular expressionof ERM proteins in normal and malignant tissues: cytoplasmic ezrin expression has prognostic signficance for head and neck squamous cell carcinoma.
Head & neck
Madan R,Brandwein-Gensler M,Schlecht NF,Elias K,Gorbovitsky E,Belbin TJ,Mahmood R,Breining D,Qian H,Childs G,Locker J,Smith R,Haigentz M,Gunn-Moore F,Prystowsky MB
MA5-14043 was used in immunohistochemistry to examine the prognostic significance of cytoplasmic ezrin expression for head and neck squamous cell carcinoma
Wed Nov 01 00:00:00 EST 2006
Ezrin promotes ovarian carcinoma cell invasion and its retained expression predicts poor prognosis in ovarian carcinoma.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Köbel M,Gradhand E,Zeng K,Schmitt WD,Kriese K,Lantzsch T,Wolters M,Dittmer J,Strauss HG,Thomssen C,Hauptmann S
MA5-14043 was used in immunohistochemistry to study the role of ezrin in ovarian carcinoma cell invasion
Sat Apr 01 00:00:00 EST 2006
Ezrin is essential for epithelial organization and villus morphogenesis in the developing intestine.
Developmental cell
Saotome I,Curto M,McClatchey AI
MA5-14043 was used in immunohistochemistry to study the essential role of Ezrin in epithelial organization and villus morphogenesis during intestinal development
Tue Jun 01 00:00:00 EDT 2004
Chemokine stimulation of human peripheral blood T lymphocytes induces rapid dephosphorylation of ERM proteins, which facilitates loss of microvilli and polarization.
Blood
Brown MJ,Nijhara R,Hallam JA,Gignac M,Yamada KM,Erlandsen SL,Delon J,Kruhlak M,Shaw S
MA5-14043 was used in immunocytochemistry to investigate the regulation of chemokines on Carboxyphosphorylated-ERM and its effect on the morphology and activation of human peripheral blood T lymphocytes
Mon Dec 01 00:00:00 EST 2003
Ezrin and moesin expression within the developing human cerebrum and tuberous sclerosis-associated cortical tubers.
Acta neuropathologica
Johnson MW,Miyata H,Vinters HV
MA5-14043 was used in immunohistochemistry to study the expression of ezrin and moesin in developing human cerebrum and tuberous sclerosis-associated cortical tubers
Thu Aug 01 00:00:00 EDT 2002
Molecular profiling of bladder cancer using cDNA microarrays: defining histogenesis and biological phenotypes.
Cancer research
Sanchez-Carbayo M,Socci ND,Charytonowicz E,Lu M,Prystowsky M,Childs G,Cordon-Cardo C
MA5-14043 was used in immunohistochemistry to study the molecular profile of bladder cancer using cDNA microarrays
Sun Dec 01 00:00:00 EST 2002