Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • N6AMT1 Antibodies

          Invitrogen

          N6AMT1 Polyclonal Antibody

          2 Published Figures
          3 References
          View all (6) N6AMT1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite N6AMT1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (2)
          N6AMT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          N6AMT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          N6AMT1 Antibody (PA5-121076) in ICC/IF

          Immunofluorescence analysis of N6AMT1 in PC-12 cells. Samples were incubated with N6AMT1 Polyclonal antibody (Product # PA5-121076) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          N6AMT1 Antibody in Immunocytochemistry (ICC/IF)
          N6AMT1 Antibody in Western Blot (WB)
          N6AMT1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          N6AMT1 Antibody in Western Blot (WB)
          N6AMT1 Polyclonal Antibody

          Product Details

          PA5-121076

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          View 1 publication 1 publication

          ELISA (ELISA)

          1 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to a sequence within amino acids 1-186 of human N6AMT1/HEMK2 (NP_8774263)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.92 mg/mL

          Amount

          192 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2914648

          Product Specific Information

          Positive test controls include: Rat kidney.

          Immunogen sequence: MAGENFATPF HGHVGRGAFS DVYEPAEDTF LLLNALEAAA AELAGVEICL EVGSGSGVVS AFLASMIGPQ ALYMCTDINP EAAACTLETA RCNKVHIQPV ITDLVGSHGI EAAWAGGRNG REVMDRFFPL VPDLLSPRGL FYLVTIKENN PEEILKIMKT KGLQGTTALS RQAGQETLSV LKFTKS

          Target Information

          Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on 'Gln-185', using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HemK methyltransferase family member 2; Lysine N-methyltransferase 9; M-N6DNA MTase A; M.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase HEMK2; Methyltransferase N6AMT1; N(6)-adenine-specific DNA methyltransferase 1; N-6 adenine-specific DNA methyltransferase 1 (putative); N6-DNA methyltransferase A; N6-DNA-methyltransferase; Protein N(5)-glutamine methyltransferase; unnamed protein product

          View more View less

          Gene Aliases: 5830445C04Rik; C21orf127; HEMK2; KMT9; m.HsaHemK2P; MTQ2; N6AMT; N6AMT1; PRED28; PrmC; RGD1311843

          View more View less

          UniProt ID: (Human) Q9Y5N5, (Mouse) Q6SKR2

          View more View less

          Entrez Gene ID: (Human) 29104, (Mouse) 67768, (Rat) 288309

          View more View less

          Function(s)
          nucleic acid binding protein binding methyltransferase activity protein methyltransferase activity S-adenosylmethionine-dependent methyltransferase activity site-specific DNA-methyltransferase (adenine-specific) activity transferase activity arsenite methyltransferase activity protein-glutamine N-methyltransferase activity histone methyltransferase activity histone H4K12 methyltransferase activity S-adenosyl-L-methionine binding methyltransferase transferase metabolite interconversion enzyme
          Process(es)
          chromatin organization toxin metabolic process peptidyl-glutamine methylation arsonoacetate metabolic process positive regulation of cell growth methylation negative regulation of gene expression, epigenetic positive regulation of gene expression, epigenetic protein methylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-rlhf6:80/100.66.79.73:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline