Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NADK Antibodies

          Proteintech

          NADK Polyclonal Antibody

          6 References
          View all (7) NADK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NADK Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (18)
          NADK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NADK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NADK Antibody (15548-1-AP) in ICC/IF

          Immunofluorescent analysis of Hela cells, using NADK antibody 15548-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NADK Antibody in Immunocytochemistry (ICC/IF)
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NADK Antibody in Western Blot (WB)
          NADK Antibody in Western Blot (WB)
          NADK Antibody in Western Blot (WB)
          NADK Antibody in Western Blot (WB)
          NADK Antibody in Western Blot (WB)
          NADK Polyclonal Antibody

          Product Details

          15548-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NADK Fusion Protein Ag7906 (98-446 aa encoded by BC001709)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.29 mg/mL

          Amount

          43.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: LTWNKSPKS VLVIKKMRDA SLLQPFKELC THLMEENMIV YVEKKVLEDP AIASDESFGA VKKKFCTFRE DYDDISNQID FIICLGGDGT LLYASSLFQG SVPPVMAFHL GSLGFLTPFS FENFQSQVTQ VIEGNAAVVL RSRLKVRVVK ELRGKKTAVH NGLGEKGSQA AGLDMDVGKQ AMQYQVLNEV VIDRGPSSYL SNVDVYLDGH LITTVQGDGV IVSTPTGSTA YAAAAGASMI HPNVPAIMIT PICPHSLSFR PIVVPAGVEL KIMLSPEARN TAWVSFDGRK RQEIRHGDSI SITTSCYPLP SICVRDPVSD WFESLAQCLH WNVRKKQAHF EEEEEEEEEG (98-446 aa encoded by BC001709)

          Target Information

          NADK (Nicotinamide Adenine Dinucleotide Kinase) is an enzyme crucial in the regulation of intracellular NADP+ levels, which are necessary for metabolic processes such as fatty acid and DNA synthesis. The human NADK gene is located on chromosome 1p36.33 and encodes a protein that primarily functions in the cytoplasm. NADK catalyzes the phosphorylation of NAD+ to NADP+, playing a significant role in maintaining the cellular redox state. Structurally, human NADK forms a dimeric organization, as revealed by its crystal structure, and includes four subunits (chains A-D) with specific colors indicating different chains. The enzyme's activity is regulated post-translationally, but its pathophysiological significance in humans remains underexplored. Multiple orthologs of the NADK gene are present in various organisms, illustrating its evolutionary conservation and importance across different species.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FLJ13052; NAD kinase; Poly(P)/ATP NAD kinase; ppnk; Putative inorganic polyphosphate/ATP-NAD kinase (Poly(P)/ATP NAD kinase); unnamed protein product

          View more View less

          Gene Aliases: 4432404C02Rik; BC004012; dJ283E3.1; NADK; NADK1

          View more View less

          UniProt ID: (Human) O95544, (Mouse) P58058

          View more View less

          Entrez Gene ID: (Human) 65220, (Rat) 100125370, (Mouse) 192185

          View more View less

          Function(s)
          nucleotide binding NAD+ kinase activity protein binding ATP binding kinase activity transferase activity metal ion binding nucleotide kinase
          Process(es)
          NADP biosynthetic process phosphorylation NAD metabolic process positive regulation of insulin secretion involved in cellular response to glucose stimulus ATP metabolic process metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline