Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NDUFB2 Antibodies

          Proteintech

          NDUFB2 Polyclonal Antibody

          View all (7) NDUFB2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NDUFB2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (27)
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 27

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NDUFB2 Antibody (17614-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using (Product # 17614-1-AP) (NDUFB2 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDUFB2 Antibody in Western Blot (WB)
          NDUFB2 Antibody in Western Blot (WB)
          NDUFB2 Antibody in Western Blot (WB)
          NDUFB2 Antibody in Western Blot (WB)
          NDUFB2 Antibody in Western Blot (WB)
          NDUFB2 Polyclonal Antibody

          Product Details

          17614-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NDUFB2 Fusion Protein Ag11789 (1-95 aa encoded by BC063026)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MESHERQPL VLHEEMGECR SSLSAGGGVH IEPRYRQFPQ LTRSQVFQSE FFSGLMWFWI LWRFWHDSEE VLGHFPYPDP SQWTDEELGI PPDDED (1-95 aa encoded by BC063026)

          Target Information

          The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CI-AGGG; complex I AGGG subunit; Complex I-AGGG; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 2, mitochondrial; NADH-ubiquinone oxidoreductase AGGG subunit; NDUFB2

          View more View less

          Gene Aliases: 1810011O01Rik; 8kDa; AGGG; AI325567; CI-AGGG; NDUFB2

          View more View less

          UniProt ID: (Human) O95178, (Mouse) Q9CPU2

          View more View less

          Entrez Gene ID: (Human) 4708, (Rat) 362344, (Mouse) 68198

          View more View less

          Function(s)
          NADH dehydrogenase (ubiquinone) activity molecular_function oxidoreductase metabolite interconversion enzyme
          Process(es)
          mitochondrial electron transport, NADH to ubiquinone aerobic respiration mitochondrial ATP synthesis coupled proton transport hydrogen ion transmembrane transport biological_process transport oxidation-reduction process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline