Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAVARAGVL GVQWLQRASR NVMPLGARTA SHMTKDMFPG PYPRTPEERA AAAKKYNMRV EDYEPYPDDG MGYGDYPKLP DRSQHERDPW YSWDQPGLRL NWGEPMHWHL DMYNRNRVDT SPTPVSWHVM CMQLFGFLAF MIFMCWVGDV YPVYQPVGPK QYPYNNLYLE RGGDPSKEPE RVVHYEI (1-186 aa encoded by BC000466)
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-ASHI; complex I ASHI subunit; complex I subunit; complex I-ASHI; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa; NADH-ubiquinone oxidoreductase ASHI subunit; NADH:ubiquinone oxidoreductase ASHI subunit; NDUFB8; unnamed protein product
Gene Aliases: 2900010I05Rik; ASHI; CI-ASHI; MC1DN32
UniProt ID: (Human) O95169, (Mouse) Q9D6J5
Entrez Gene ID: (Human) 4714, (Rat) 293991, (Mouse) 67264
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support