Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody can recognize NEDD4-1 and NEDD4-2.
Immunogen sequence: EESYRRIMS VKRPDVLKAR LWIEFESEKG LDYGGVAREW FFLLSKEMFN PYYGLFEYSA TDNYTLQINP NSGLCNEDHL SYFTFIGRVA GLAVFHGKLL DGFFIRPFYK MMLGKQITLN DMESVDSEYY NSLKWILEND PTELDLMFCI DEENFGQTYQ VDLKPNGSEI MVTNENKREY IDLVIQWRFV NRVQKQMNAF LEGFTELLPI DLIKIFDENE LELLMCGLGD VDVNDWRQHS IYKNGYCPNH PVIQWFWKAV LLMDAEKRIR LLQFVTGTSR VPMNGFAELY GSNGPQLFTI EQWGSPEKLP RAHTCFNRLD LPPYETFEDL REKLLMAVEN AQGFEGVD (565-911 aa encoded by BC032597)
NEDD4L is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. NEDD4L inhibits TGF-beta signaling by triggering SMAD2 and TGFR1 ubiquitination and proteasome-dependent degradation. NEDD4L promotes ubiquitination and internalization of various plasma membrane channels such as ENaC, Nav1.2, Nav1.3, Nav1.5, Nav1.7, Nav1.8, Kv1.3, EAAT1 or CLC5. NEDD4L also promotes ubiquitination and degradation of SGK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternatively spliced; amino-terminal variant; developmentally down-regulated 4-like; FLJ33870; HECT-type E3 ubiquitin transferase NED4L; KIAA0439; neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4b; similar to NEDD4; ubiquitin-ligase; neural precursor cell expressed, developmentally down-regulated 4-like protein; ubiquitin-protein ligase Rsp5; unnamed protein product
Gene Aliases: 1300012C07Rik; hNEDD4-2; NEDD4-2; NEDD4.2; Nedd4b; PVNH7; RSP5
UniProt ID: (Human) Q96PU5
Entrez Gene ID: (Human) 23327, (Mouse) 83814, (Rat) 291553
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support