Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • NEDD4L Antibodies

          Bethyl Laboratories

          NEDD4L Polyclonal Antibody

          View all (21) NEDD4L antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NEDD4L Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NEDD4L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NEDD4L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NEDD4L Antibody (A302-512A-T) in WB

          Detection of human and mouse NEDD4L by western blot (h&m) and immunoprecipitation (h). Samples: Whole cell lysate from HeLa (5, 15 and 50 µg for WB; 1 mg for IP, 20% of IP loaded), HEK293T (T; 50 µg), and mouse NIH 3T3 (M; 50 µg) cells. Antibodies: Affinity purified rabbit anti-NEDD4L antibody (Product # A302-512A) used for WB at 0.04 µg/mL (A) and 0.4 µg/mL (B) and used for IP at 3 µg/mg lysate. NEDD4L was also immunoprecipitated by rabbit anti-NEDD4L antibodies A302-513A and A302-514A, which recognize downstream epitopes. Detection: Chem... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NEDD4L Antibody in Western Blot (WB)
          NEDD4L Polyclonal Antibody

          Product Details

          A302-512A-T

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:10,000
          -

          Immunoprecipitation (IP)

          2-5 µg/mg lysate
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Region between residue 225 and 275 of human neural precursor cell expressed, developmentally down-regulated 4-like.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.20 mg/mL

          Amount

          2 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          TBS, pH 7.0 to 8.0, with 0.1% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          The recommended shelf life for this product is 1 year from date of receipt.

          Based on 100% sequence identity, this antibody is predicted to react with Orangutan

          Target Information

          NEDD4L is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. NEDD4L inhibits TGF-beta signaling by triggering SMAD2 and TGFR1 ubiquitination and proteasome-dependent degradation. NEDD4L promotes ubiquitination and internalization of various plasma membrane channels such as ENaC, Nav1.2, Nav1.3, Nav1.5, Nav1.7, Nav1.8, Kv1.3, EAAT1 or CLC5. NEDD4L also promotes ubiquitination and degradation of SGK.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alternatively spliced; amino-terminal variant; developmentally down-regulated 4-like; E3 ubiquitin-protein ligase NEDD4-like; FLJ33870; HECT-type E3 ubiquitin transferase NED4L; KIAA0439; Nedd4-2; NEDD4.2; neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4b; similar to NEDD4; ubiquitin-ligase; neural precursor cell expressed, developmentally down-regulated 4-like protein; ubiquitin-protein ligase Rsp5; unnamed protein product

          View more View less

          Gene Aliases: 1300012C07Rik; hNEDD4-2; KIAA0439; NEDD4-2; NEDD4.2; Nedd4b; NEDD4L; NEDL3; PVNH7; RSP5

          View more View less

          UniProt ID: (Human) Q96PU5

          View more View less

          Entrez Gene ID: (Human) 23327, (Mouse) 83814

          View more View less

          Function(s)
          ubiquitin-protein transferase activity protein binding transferase activity sodium channel regulator activity potassium channel inhibitor activity sodium channel inhibitor activity ion channel binding ubiquitin protein ligase activity UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity potassium channel regulator activity ligase activity ribosomal S6-glutamic acid ligase activity coenzyme F420-0 gamma-glutamyl ligase activity coenzyme F420-2 alpha-glutamyl ligase activity protein-glycine ligase activity protein-glycine ligase activity, initiating protein-glycine ligase activity, elongating tubulin-glycine ligase activity protein-glutamic acid ligase activity tubulin-glutamic acid ligase activity ubiquitin-protein ligase
          Process(es)
          regulation of membrane depolarization ubiquitin-dependent protein catabolic process protein monoubiquitination ion transport neuromuscular junction development protein ubiquitination cell differentiation neuron projection development receptor internalization regulation of protein stability receptor catabolic process ion transmembrane transport regulation of membrane potential proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of protein catabolic process regulation of dendrite morphogenesis regulation of synapse organization regulation of membrane repolarization regulation of biological quality protein K48-linked ubiquitination ventricular cardiac muscle cell action potential sodium ion import across plasma membrane negative regulation of potassium ion transmembrane transport regulation of sodium ion transmembrane transport negative regulation of sodium ion transmembrane transport negative regulation of potassium ion export across plasma membrane negative regulation of sodium ion import across plasma membrane positive regulation of dendrite extension negative regulation of protein localization to cell surface positive regulation of caveolin-mediated endocytosis negative regulation of systemic arterial blood pressure response to salt stress response to metal ion positive regulation of sodium ion transport negative regulation of sodium ion transport regulation of ion transmembrane transport protein ubiquitination involved in ubiquitin-dependent protein catabolic process ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway regulation of potassium ion transmembrane transporter activity negative regulation of potassium ion transmembrane transporter activity negative regulation of sodium ion transmembrane transporter activity regulation of bicellular tight junction assembly positive regulation of cation channel activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline