Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • NELL1 Antibodies

          Abnova

          NELL1 Polyclonal Antibody, MaxPab™

          View all (11) NELL1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NELL1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          NELL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NELL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NELL1 Antibody (H00004745-B01P) in WB

          Western Blot analysis of NELL1 expression in transfected 293T cell line (H00004745-T01) by NELL1 MaxPab polyclonal antibody.

          Lane 1: NELL1 transfected lysate(89.1 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NELL1 Antibody in Western Blot (WB)
          NELL1 Antibody in Western Blot (WB)
          NELL1 Polyclonal Antibody, MaxPab™

          Product Details

          H00004745-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NELL1 (AAH69674.1, 1 a.a. approximately 810 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MPMDLILVVW FCVCTARTVV GFGMDPDLQM DIVTELDLVN TTLGVAQVSG MHNASKAFLF QDIEREIHAA PHVSEKLIQL FRNKSEFTIL ATVQQKPSTS GVILSIRELE HSYFELESSG LRDEIRYHYI HNGKPRTEAL PYRMADGQWH KVALSVSASH LLLHVDCNRI YERVIDPPDT NLPPGINLWL GQRNQKHGLF KGIIQDGKII FMPNGYITQC PNLNHTCPTC SDFLSLVQGI MDLQELLAKM TAKLNYAETR LSQLENCHCE KTCQVSGLLY RDQDSWVDGD HCRNCTCKSG AVECRRMSCP PLNCSPDSLP VHIAGQCCKV CRPKCIYGGK VLAEGQRILT KSCRECRGGV LVKITEMCPP LNCSEKDHIL PENQCCRVCR GHNFCAEGPK CGENSECKNW NTKATCECKS GYISVQGDSA YCEDIDECAA KMHYCHANTV CVNLPGLYRC DCVPGYIRVD DFSCTEHDEC GSGQHNCDEN AICTNTVQGH SCTCKPGYVG NGTICRAFCE EGCRYGGTCV APNKCVCPSG FTGSHCEKDI DECSEGIIEC HNHSRCVNLP GWYHCECRSG FHDDGTYSLS GESCIDIDEC ALRTHTCWND SACINLAGGF DCLCPSGPSC SGDCPHEGGL KHNGQVWTLK EDRCSVCSCK DGKIFCRRTA CDCQNPSADL FCCPECDTRV TSQCLDQNGH KLYRSGDNWT HSCQQCRCLE GEVDCWPLTC PNLSCEYTAI LEGECCPRCV SDPCLADNIT YDIRKTCLDS YGVSRLSGSV WTMAGSPCTT CKCKNGRVCC SVDFECLQNN

          Target Information

          This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: nel homolog; NEL-like protein 1; Nel-related protein 1; neural epidermal growth factor-like 1; Protein kinase C-binding protein NELL1; unnamed protein product

          View more View less

          Gene Aliases: IDH3GL; NELL1; NRP1

          View more View less

          UniProt ID: (Human) Q92832

          View more View less

          Entrez Gene ID: (Human) 4745

          View more View less

          Function(s)
          protein kinase C binding calcium ion binding protein binding heparin binding
          Process(es)
          nervous system development regulation of gene expression cell differentiation positive regulation of bone mineralization negative regulation of osteoblast proliferation negative regulation of protein catabolic process regulation of osteoblast differentiation positive regulation of osteoblast differentiation positive regulation of ossification
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline