Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • NFAT5 Antibodies

          Abnova

          NFAT5 Polyclonal Antibody, MaxPab™

          View all (11) NFAT5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NFAT5 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NFAT5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NFAT5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NFAT5 Antibody (H00010725-B01P) in WB

          Western Blot analysis of NFAT5 expression in transfected 293T cell line by NFAT5 MaxPab polyclonal antibody. Lane 1: NFAT5 transfected lysate(163.24 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NFAT5 Antibody in Western Blot (WB)

          Product Details

          H00010725-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NFAT5 (NP_619727.1, 1 a.a. approximately 1484 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSQTSGGEAG SPPPAVVAAD ASSAPSSSSM GGACSSFTTS SSPTIYSTSV TDSKAMQVES CSSAVGVSNR GVSEKQLTSN TVQQHPSTPK RHTVLYISPP PEDLLDNSRM SCQDEGCGLE SEQSCSMWME DSPSNFSNMS TSSYNDNTEV PRKSRKRNPK QRPGVKRRDC EESNMDIFDA DSAKAPHYVL SQLTTDNKGN SKAGNGTLEN QKGTGVKKSP MLCGQYPVKS EGKELKIVVQ PETQHRARYL TEGSRGSVKD RTQQGFPTVK LEGHNEPVVL QVFVGNDSGR VKPHGFYQAC RVTGRNTTPC KEVDIEGTTV IEVGLDPSNN MTLAVDCVGI LKLRNADVEA RIGIAGSKKK STRARLVFRV NIMRKDGSTL TLQTPSSPIL CTQPAGVPEI LKKSLHSCSV KGEEEVFLIG KNFLKGTKVI FQENVSDENS WKSEAEIDME LFHQNHLIVK VPPYHDQHIT LPVSVGIYVV TNAGRSHDVQ PFTYTPDPAA AGALNVNVKK EISSPARPCS FEEAMKAMKT TGCNLDKVNI IPNALMTPLI PSSMIKSEDV TPMEVTAEKR SSTIFKTTKS VGSTQQTLEN ISNIAGNGSF SSPSSSHLPS ENEKQQQIQP KAYNPETLTT IQTQDISQPG TFPAVSASSQ LPNSDALLQQ ATQFQTRETQ SREILQSDGT VVNLSQLTEA SQQQQQSPLQ EQAQTLQQQI SSNIFPSPNS VSQLQNTIQQ LQAGSFTGST ASGSSGSVDL VQQVLEAQQQ LSSVLFSAPD GNENVQEQLS ADIFQQVSQI QSGVSPGMFS STEPTVHTRP DNLLPGRAES VHPQSENTLS NQQQQQQQQQ QVMESSAAMV MEMQQSICQA AAQIQSELFP STASANGNLQ QSPVYQQTSH MMSALSTNED MQMQCELFSS PPAVSGNETS TTTTQQVATP GTTMFQTSSS GDGEETGTQA KQIQNSVFQT MVQMQHSGDN QPQVNLFSST KSMMSVQNSG TQQQGNGLFQ QGNEMMSLQS GNFLQQSSHS QAQLFHPQNP IADAQNLSQE TQGSLFHSPN PIVHSQTSTT SSEQMQPPMF HSQSTIAVLQ GSSVPQDQQS TNIFLSQSPM NNLQTNTVAQ EAFFAAPNSI SPLQSTSNSE QQAAFQQQAP ISHIQTPMLS QEQAQPPQQG LFQPQVALGS LPPNPMPQSQ QGTMFQSQHS IVAMQSNSPS QEQQQQQQQQ QQQQQQQQQS ILFSNQNTMA TMASPKQPPP NMIFNPNQNP MANQEQQNQS IFHQQSNMAP MNQEQQPMQF QSQSTVSSLQ NPGPTQSESS QTPLFHSSPQ IQLVQGSPSS QEQQVTLFLS PASMSALQTS INQQDMQQSP LYSPQNNMPG IQGATSSPQP QATLFHNTAG GTMNQLQNSP GSSQQTSGMF LFGIQNNCSQ LLTSGPATLP DQLMAISQPG QPQNEGQPPV TTLLSQQMPE NSPLASSINT NQNIEKIDLL VSLQNQGNNL TGSF

          Target Information

          The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: glutamine rich protein H65; NF-AT5; NFAT-like protein 1; NFAT5b; Nuclear factor of activated T-cells 5; nuclear factor of activated T-cells 5, tonicity-responsive; osmotic response element-binding protein; T-cell transcription factor NFAT5; TonE-binding protein; TonEBP; transcription factor; Tonicity-responsive enhancer-binding protein

          View more View less

          Gene Aliases: KIAA0827; NF-AT5; NFAT5; NFATL1; NFATZ; OREBP; TONEBP

          View more View less

          UniProt ID: (Human) O94916

          View more View less

          Entrez Gene ID: (Human) 10725

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific double-stranded DNA binding
          Process(es)
          transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter response to stress cellular response to DNA damage stimulus signal transduction positive regulation of gene expression calcineurin-NFAT signaling cascade positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of transcription from RNA polymerase II promoter R-loop processing regulation of calcineurin-NFAT signaling cascade cellular response to cytokine stimulus cellular hyperosmotic response regulation of intracellular signal transduction positive regulation of leukocyte adhesion to vascular endothelial cell
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline