Product References
Combined blockade of the IL-13 and IL-33 pathways leads to a greater inhibition of type 2 inflammation over inhibition of either pathway alone.
The Journal of allergy and clinical immunology
Ramirez-Carrozzi V,Sambandam A,Zhou M,Yan D,Kang J,Wu X,Suto E,Baca M,Austin C,Xu M,Lee W,Pappu R
MA516916 was used in flow cytometry to study the effect of IL-13 and IL-33 pathways in dysregulated type 2 inflammation.
Wed Feb 01 00:00:00 EST 2017
Glucocorticoid receptor in T cells mediates protection from autoimmunity in pregnancy.
Proceedings of the National Academy of Sciences of the United States of America
Engler JB,Kursawe N,Solano ME,Patas K,Wehrmann S,Heckmann N,Lühder F,Reichardt HM,Arck PC,Gold SM,Friese MA
MA516916 was used in flow cytometry to characterize conventional and regulatory T cells during pregnancy
Tue Jan 10 00:00:00 EST 2017
Astrocyte-derived interleukin-15 exacerbates ischemic brain injury via propagation of cellular immunity.
Proceedings of the National Academy of Sciences of the United States of America
Li M,Li Z,Yao Y,Jin WN,Wood K,Liu Q,Shi FD,Hao J
MA516916 was used in flow cytometry to discuss the contribution of astrocytic IL-15 to postischemic brain damage
Tue Jan 17 00:00:00 EST 2017
NK cells regulate CXCR2+ neutrophil recruitment during acute lung injury.
Journal of leukocyte biology
Hoegl S,Ehrentraut H,Brodsky KS,Victorino F,Golden-Mason L,Eltzschig HK,McNamee EN
MA516916 was used in flow cytometry to elucidate the role of Tbet in acute lung injury
Wed Feb 01 00:00:00 EST 2017
ATP Binding Cassette Transporter ABCA7 Regulates NKT Cell Development and Function by Controlling CD1d Expression and Lipid Raft Content.
Scientific reports
Nowyhed HN,Chandra S,Kiosses W,Marcovecchio P,Andary F,Zhao M,Fitzgerald ML,Kronenberg M,Hedrick CC
MA516916 was used in flow cytometry to show that ABCA7 regulates natural killer T cell development in a cell-extrinsic manner
Mon Jan 16 00:00:00 EST 2017
Smad3 promotes cancer progression by inhibiting E4BP4-mediated NK cell development.
Nature communications
Tang PM,Zhou S,Meng XM,Wang QM,Li CJ,Lian GY,Huang XR,Tang YJ,Guan XY,Yan BP,To KF,Lan HY
MA516916 was used in flow cytometry to demonstrate that Smad3 in the tumor microenvironment regulates cancer progression
Mon Mar 06 00:00:00 EST 2017
Targeted antibody-mediated depletion of murine CD19 CAR T cells permanently reverses B cell aplasia.
The Journal of clinical investigation
Paszkiewicz PJ,Fräßle SP,Srivastava S,Sommermeyer D,Hudecek M,Drexler I,Sadelain M,Liu L,Jensen MC,Riddell SR,Busch DH
MA516916 was used in flow cytometry to demonstrate that targeting a truncated version of the epidermal growth factor receptor with cetuximab eliminates CD19 chimeric antigen receptor T cells
Tue Nov 01 00:00:00 EDT 2016
Carbonic anhydrase enzymes regulate mast cell-mediated inflammation.
The Journal of experimental medicine
Henry EK,Sy CB,Inclan-Rico JM,Espinosa V,Ghanny SS,Dwyer DF,Soteropoulos P,Rivera A,Siracusa MC
MA516916 was used in flow cytometry to find a role for Car enzymes in regulating mast cell lineage commitment
Mon Aug 22 00:00:00 EDT 2016
Differential Role of Hematopoietic and Nonhematopoietic Cell Types in the Regulation of NK Cell Tolerance and Responsiveness.
Journal of immunology (Baltimore, Md. : 1950)
Shifrin NT,Kissiov DU,Ardolino M,Joncker NT,Raulet DH
MA516916 was used in flow cytometry to study the role of MCH I in natural killer cell tolerance and hyporesponsiveness
Tue Nov 15 00:00:00 EST 2016
CXCL16-positive dendritic cells enhance invariant natural killer T cell-dependent IFNγ production and tumor control.
Oncoimmunology
Veinotte L,Gebremeskel S,Johnston B
MA516916 was used in flow cytometry to study the contribution of CXCR6/CXCL16 interactions in glycolipid-dependent invariant natural killer cell activation and tumor control
Wed Jun 01 00:00:00 EDT 2016
DNAM-1 controls NK cell activation via an ITT-like motif.
The Journal of experimental medicine
Zhang Z,Wu N,Lu Y,Davidson D,Colonna M,Veillette A
MA5-16916 was used in flow cytometry to study how DNAM-1 controls NK cell-mediated cytotoxicity and cytokine production
Mon Nov 16 00:00:00 EST 2015
A novel Cd8-cis-regulatory element preferentially directs expression in CD44hiCD62L+ CD8+ T cells and in CD8αα+ dendritic cells.
Journal of leukocyte biology
Sakaguchi S,Hombauer M,Hassan H,Tanaka H,Yasmin N,Naoe Y,Bilic I,Moser MA,Hainberger D,Mayer H,Seiser C,Bergthaler A,Taniuchi I,Ellmeier W
MA5-16916 was used in ChIP assay to identify Cd8 cis-regulatory elements
Wed Apr 01 00:00:00 EDT 2015
Ectopic TBX1 suppresses thymic epithelial cell differentiation and proliferation during thymus organogenesis.
Development (Cambridge, England)
Reeh KA,Cardenas KT,Bain VE,Liu Z,Laurent M,Manley NR,Richie ER
MA5-16916 was used in flow cytometry to assess the contribution of Tbx1 to thymus and parathyroid development
Fri Aug 01 00:00:00 EDT 2014
Long-term, multilineage hematopoiesis occurs in the combined absence of beta-catenin and gamma-catenin.
Blood
Jeannet G,Scheller M,Scarpellino L,Duboux S,Gardiol N,Back J,Kuttler F,Malanchi I,Birchmeier W,Leutz A,Huelsken J,Held W
MA5-16916 was used in flow cytometry to investigate whether hematopoietic cells transduce canonical Wnt signals in the absence of beta- and gamma-catenin
Tue Jan 01 00:00:00 EST 2008
Thymic alterations in mice deficient for the SNARE protein VAMP8/endobrevin.
Cell and tissue research
Kanwar N,Fayyazi A,Backofen B,Nitsche M,Dressel R,von Mollard GF
MA5-16916 was used in flow cytometry to elucidate the role of VAMP8 in the thymus
Sat Nov 01 00:00:00 EDT 2008
T-bet concomitantly controls migration, survival, and effector functions during the development of Valpha14i NKT cells.
Blood
Matsuda JL,Zhang Q,Ndonye R,Richardson SK,Howell AR,Gapin L
MA5-16916 was used in flow cytometry to examine the role of T-bet in Valpha14i natural killer T cell function
Sat Apr 01 00:00:00 EST 2006
A comparison of murine T-cell-depleted adult bone marrow and full-term fetal blood cells in hematopoietic engraftment and immune reconstitution.
Blood
Chen BJ,Cui X,Sempowski GD,Gooding ME,Liu C,Haynes BF,Chao NJ
MA5-16916 was used in flow cytometry to compare myeloid and lymphoid recovery using bone marrow and cord blood as sources of stem cells.
Tue Jan 01 00:00:00 EST 2002
MHC class I molecules on adenovirus E1A-expressing tumor cells inhibit NK cell killing but not NK cell-mediated tumor rejection.
International immunology
Routes JM,Ryan JC,Ryan S,Nakamura M
MA5-16916 was used in flow cytometry to investigate the capacity of E1A to sensitize cells to multiple NK cell killing mechanisms
Mon Oct 01 00:00:00 EDT 2001
Mouse NKR-P1B, a novel NK1.1 antigen with inhibitory function.
Journal of immunology (Baltimore, Md. : 1950)
Carlyle JR,Martin A,Mehra A,Attisano L,Tsui FW,Zúñiga-Pflücker JC
MA5-16916 was used in flow cytometry and immunoprecipitation to identify NKR-P1B as a NK1.1 antigen
Sat May 15 00:00:00 EDT 1999
Natural killer cell proliferation induced by anti-NK1.1 and IL-2.
Immunology and cell biology
Reichlin A,Yokoyama WM
MA5-16916 was used in flow cytometry to report that immobilized anti-NK1.1 triggers only a subpopulation of NK cells to kill
Wed Apr 01 00:00:00 EST 1998
Pan natural killer cell monoclonal antibodies and their relationship to the NK1.1 antigen.
Hybridoma
Sentman CL,Hackett J,Moore TA,Tutt MM,Bennett M,Kumar V
MA5-16916 was used in flow cytometry to describe three new NK specific monoclonal antibodies
Fri Dec 01 00:00:00 EST 1989