Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MPLDPYHSV TWGHAQGSHH FVFGELRVRP ILESLRDEPD PDPRPSREGP AGRVGALARG GPEPCDAASP PGGASCAPEL ARPREDKSAQ QAKLEGGTRL CCRCPEESRL VPGGAVSPGD HVLEVSGTRG TCGCRPRRHA GPELAHS (1-146 aa encoded by BC016965)
NALP1 is a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apaf-1/Ced-4 family apoptosis regulator; CARD7; caspase recruitment domain protein; caspase recruitment domain protein 7; caspase recruitment domain-containing protein 7; contains NACHT, LRR and PYD domains; death effector filament-forming Ced-4-like apoptosis protein; DKFZp586O1822; nacht; NACHT, leucine rich repeat and PYD (pyrin domain) containing 1; NACHT, leucine rich repeat and PYD containing 1; NACHT, LRR and PYD containing protein 1; NACHT/LRR/pyrin domain-containing protein 1; NLR family, pyrin domain containing 1; NLR family, pyrin domain containing 1A, CAST/EiJ strain specific; NLR family, pyrin domain containing 1A, PWK/PhJ strain specific; NLR family, pyrin domain containing 1A, WSB/EiJ strain specific; NOD-LRR protein; nucleotide-binding domain and caspase recruitment domain; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 1; unnamed protein product
Gene Aliases: AIADK; CARD7; CIDED; CLR17.1; DEFCAP; DEFCAP-L/S; Gm14; Gm15; JRRP; MSPC; NAC; NALP1; Nalp1a; Nlrp1; PP1044; SLEV1; VAMAS1
UniProt ID: (Human) Q9C000, (Mouse) Q2LKU9
Entrez Gene ID: (Human) 22861, (Mouse) 195046
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support