Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGGAWGRLA CYLEFLKKEE LKEFQLLLAN KAHSRSSSGE TPAQPEKTSG MEVASYLVAQ YGEQRAWDLA LHTWEQMGLR SLCAQAQEGA GHSPSFPYSP SEPHLGSPSQ PTSTAVLMPW IHELPAGCTQ GSERRVLRQL PDTSGRRWRE ISASLLYQAL PSSPDHESPS QESPNAPTST AVLGSWGSPP QPSLAPREQE APGTQWPLDE TSGIYYTEIR EREREKSEKG RPPWAAVVGT PPQAHTSLQP HHHPWEPSVR ESLCSTWPWK NEDFNQKFTQ LLLLQRPHPR SQDPLVKRSW PDYVEENRGH LIEIRDLFGP GLDTQEPRIV ILQGAAGIGK STLARQVKEA WGRGQLYGDR FQHVFYFSCR ELAQSKVVSL AELIGKDGTA TPAPIRQILS RPERLLFILD GVDEPGWVLQ EPSSELCLHW SQPQPADALL GSLLGKTILP EASFLITART TALQNLIPSL EQARWVEVLG FSESSRKEYF YRYFTDERQA IRAFRLVKSN KELWALCLVP WVSWLACTCL MQQMKRKEKL TLTSKTTTTL CLHYLAQALQ AQPLGPQLRD LCSLAAEGIW QKKTLFSPDD LRKHGLDGAI ISTFLKMGIL QEHPIPLSYS FIHLCFQEFF AAMSYVLEDE KGRGKHSNCI IDLEKTLEAY GIHGLFGAST TRFLLGLLSD EGEREMENIF HCRLSQGRNL MQWVPSLQLL LQPHSLESLH CLYETRNKTF LTQVMAHFEE MGMCVETDME LLVCTFCIKF SRHVKKLQLI EGRQHRSTWS PTMVVLFRWV PVTDAYWQIL FSVLKVTRNL KELDLSGNSL SHSAVKSLCK TLRRPRCLLE TLRLAGCGLT AEDCKDLAFG LRANQTLTEL DLSFNVLTDA GAKHLCQRLR QPSCKLQRLQ LVSCGLTSDC CQDLASVLSA SPSLKELDLQ QNNLDDVGVR LLCEGLRHPA CKLIRLGLDQ TTLSDEMRQE LRALEQEKPQ LLIFSRRKPS VMTPTEGLDT GEMSNSTSSL KRQRLGSERA ASHVAQANLK LLDVSKIFPI AEIAEESSPE VVPVELLCVP SPASQGDLHT KPLGTDDDFW GPTGPVATEV VDKEKNLYRV HFPVAGSYRW PNTGLCFVMR EAVTVEIEFC VWDQFLGEIN PQHSWMVAGP LLDIKAEPGA VEAVHLPHFV ALQGGHVDTS LFQMAHFKEE GMLLEKPARV ELHHIVLENP SFSPLGVLLK MIHNALRFIP VTSVVLLYHR VHPEEVTFHL YLIPSDCSIR KELELCYRSP GEDQLFSEFY VGHLGSGIRL QVKDKKDETL VWEALVKPGD LMPATTLIPP ARIAVPSPLD APQLLHFVDQ YREQLIARVT SVEVVLDKLH GQVLSQEQYE RVLAENTRPS QMRKLFSLSQ SWDRKCKDGL YQALKETHPH LIMELWEKGS KKGLLPLSS
NALP1 is a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apaf-1/Ced-4 family apoptosis regulator; CARD7; caspase recruitment domain protein 7; Caspase recruitment domain-containing protein 7; contains NACHT, LRR and PYD domains; Death effector filament-forming ced-4-like apoptosis protein; DKFZp586O1822; nacht; NACHT, leucine rich repeat and PYD (pyrin domain) containing 1; NACHT, leucine rich repeat and PYD containing 1; NACHT, LRR and PYD containing protein 1; NACHT, LRR and PYD domains-containing protein 1; Nucleotide-binding domain and caspase recruitment domain; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 1; unnamed protein product
Gene Aliases: AIADK; CARD7; CIDED; CLR17.1; DEFCAP; DEFCAP-L/S; JRRP; KIAA0926; MSPC; NAC; NALP1; NLRP1; PP1044; SLEV1; VAMAS1
UniProt ID: (Human) Q9C000
Entrez Gene ID: (Human) 22861
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support