Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • NALP11 Antibodies

          Abnova

          NLRP11 Polyclonal Antibody, MaxPab™

          View all (6) NALP11 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NLRP11 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NLRP11 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NLRP11 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NLRP11 Antibody (H00204801-B01P) in WB

          Western Blot analysis of NLRP11 expression in transfected 293T cell line by NLRP11 MaxPab polyclonal antibody. Lane 1: NALP11 transfected lysate(113.63 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NLRP11 Antibody in Western Blot (WB)

          Product Details

          H00204801-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NLRP11 (NP_659444.1, 1 a.a. approximately 1033 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAESDSTDFD LLWYLENLSD KEFQSFKKYL ARKILDFKLP QFPLIQMTKE ELANVLPISY EGQYIWNMLF SIFSMMRKED LCRKIIGRRN RNQEACKAVM RRKFMLQWES HTFGKFHYKF FRDVSSDVFY ILQLAYDSTS YYSANNLNVF LMGERASGKT IVINLAVLRW IKGEMWQNMI SYVVHLTSHE INQMTNSSLA ELIAKDWPDG QAPIADILSD PKKLLFILED LDNIRFELNV NESALCSNST QKVPIPVLLV SLLKRKMAPG CWFLISSRPT RGNNVKTFLK EVDCCTTLQL SNGKREIYFN SFFKDRQRAS AALQLVHEDE ILVGLCRVAI LCWITCTVLK RQMDKGRDFQ LCCQTPTDLH AHFLADALTS EAGLTANQYH LGLLKRLCLL AAGGLFLSTL NFSGEDLRCV GFTEADVSVL QAANILLPSN THKDRYKFIH LNVQEFCTAI AFLMAVPNYL IPSGSREYKE KREQYSDFNQ VFTFIFGLLN ANRRKILETS FGYQLPMVDS FKWYSVGYMK HLDRDPEKLT HHMPLFYCLY ENREEEFVKT IVDALMEVTV YLQSDKDMMV SLYCLDYCCH LRTLKLSVQR IFQNKEPLIR PTASQMKSLV YWREICSLFY TMESLRELHI FDNDLNGISE RILSKALEHS SCKLRTLKLS YVSTASGFED LLKALARNRS LTYLSINCTS ISLNMFSLLH DILHEPTCQI SHLSLMKCDL RASECEEIAS LLISGGSLRK LTLSSNPLRS DGMNILCDAL LHPNCTLISL VLVFCCLTEN CCSALGRVLL FSPTLRQLDL CVNRLKNYGV LHVTFPLLFP TCQLEELHLS GCFFSSDICQ YIAIVIATNE KLRSLEIGSN KIEDAGMQLL CGGLRHPNCM LVNIGLEECM LTSACCRSLA SVLTTNKTLE RLNLLQNHLG NDGVAKLLES LISPDCVLKV VGLPLTGLNT QTQQLLMTVK ERKPSLIFLS ETWSLKEGRE IGVTPASQPG SIIPNSNLDY MFFKFPRMSA AMRTSNTASR QPL

          Target Information

          NALP11 is a 1033 amino acid containing cytoplasmic protein belonging to the NLRP family with a DAPIN domain, six LRR (leucine-rich) repeats and a NACHT domain. Reports suggest that it is a short NALP involved in inflammation potential. NALP proteins are implicated in the activation of proinflammatory caspases (e.g., CASP1) via their involvement in multiprotein complexes called inflammasomes. The DAPIN domain is necessary and sufficient for suppression of NF-kappa B activation induced by TNF and for inducing IL-1B secretion in collaboration with Caspase-1. It is involved in interaction with PYCARD.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: NACHT, leucine rich repeat and PYD containing 11; NACHT, LRR and PYD domains-containing protein 11; NOD17; Nucleotide-binding oligomerization domain protein 17; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 11; PAAD- and NACHT-containing protein 10B; PAAD-and NACHT domain-containing protein 10; PYRIN-containing APAF1-like protein 6; unnamed protein product

          View more View less

          Gene Aliases: CLR19.6; NALP11; NLRP11; NOD17; PAN10; PYPAF6; PYPAF7

          View more View less

          UniProt ID: (Human) P59045

          View more View less

          Entrez Gene ID: (Human) 204801

          View more View less

          Function(s)
          nucleotide binding RNA binding ATP binding oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced ascorbate as one donor, and incorporation of one atom of oxygen
          Process(es)
          regulation of inflammatory response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline