Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • NOD2 Antibodies

          Abnova

          NOD2 Polyclonal Antibody, MaxPab™

          View all (15) NOD2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NOD2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NOD2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NOD2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NOD2 Antibody (H00064127-D01P) in WB

          Western Blot analysis of NOD2 expression in transfected 293T cell line (H00064127-T01) by NOD2 MaxPab polyclonal antibody.

          Lane 1: NOD2 transfected lysate(114.40 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NOD2 Antibody in Western Blot (WB)
          NOD2 Polyclonal Antibody, MaxPab™

          Product Details

          H00064127-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NOD2 (AAI56572.1, 1 a.a. approximately 1040 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MGEEGGSASH DEEERASVLL GHSPGCEMCS QEAFQAQRSQ LVELLVSGSL EGFESVLDWL LSWEVLSWED YEGFHLLGQP LSHLARRLLD TVWNKGTWAC QKLIAAAQEA QADSQSPKLH GCWDPHSLHP ARDLQSHRPA IVRRLHSHVE NMLDLAWERG FVSQYECDEI RLPIFTPSQR ARRLLDLATV KANGLAAFLL QHVQELPVPL ALPLEAATCK KYMAKLRTTV SAQSRFLSTY DGAETLCLED IYTENVLEVW ADVGMAGPPQ KSPATLGLEE LFSTPGHLND DADTVLVVGE AGSGKSTLLQ RLHLLWAAGQ DFQEFLFVFP FSCRQLQCMA KPLSVRTLLF EHCCWPDVGQ EDIFQLLLDH PDRVLLTFDG FDEFKFRFTD RERHCSPTDP TSVQTLLFNL LQGNLLKNAR KVVTSRPAAV SAFLRKYIRT EFNLKGFSEQ GIELYLRKRH HEPGVADRLI RLLQETSALH GLCHLPVFSW MVSKCHQELL LQEGGSPKTT TDMYLLILQH FLLHATPPDS ASQGLGPSLL RGRLPTLLHL GRLALWGLGM CCYVFSAQQL QAAQVSPDDI SLGFLVRAKG VVPGSTAPLE FLHITFQCFF AAFYLALSAD VPPALLRHLF NCGRPGNSPM ARLLPTMCIQ ASEGKDSSVA ALLQKAEPHN LQITAAFLAG LLSREHWGLL AECQTSEKAL LRRQACARWC LARSLRKHFH SIPPAAPGEA KSVHAMPGFI WLIRSLYEMQ EERLARKAAR GLNVGHLKLT FCSVGPTECA ALAFVLQHLR RPVALQLDYN SVGDIGVEQL LPCLGVCKAL YLRDNNISDR GICKLIECAL HCEQLQKLAL FNNKLTDGCA HSMAKLLACR QNFLALRLGN NYITAAGAQV LAEGLRGNTS LQFLGFWGNR VGDEGAQALA EALGDHQSLR WLSLVGNNIG SVGAQALALM LAKNVMLEEL CLEENHLQDE GVCSLAEGLK KNSSLKILKL SNNCITYLGA EALLQALERN DTILEVWLRG NTFSLEEVDK LGCRDTRLLL

          Target Information

          NOD2 is a member of the apoptosis regulating protein family that includes caspase recruitment-domains, as well as Apaf-1 and NOD1. It contains two N-terminal CARDs, a nucleotide binding domain (NBD), and multiple C-terminal leucine-rich repeats (LRRs). NOD2 is expressed in monocytes (whereas NOD1 is expressed in multiple tissues). NOD2 plays a role in regulating NF-kappaB, acts as an intracellular receptor for bacterial lipopolysaccharides, and contributes to inflammatory bowel disease (IBD) and Crohn's disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: caspase recruitment domain family, member 15; caspase recruitment domain protein 15; Caspase recruitment domain-containing protein 15; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; NOD 2; NOD-2; NOD-like receptor C2; NOD2 protein; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; Nucleotide-binding oligomerization domain-containing protein 2; unnamed protein product

          View more View less

          Gene Aliases: ACUG; BLAU; BLAUS; CARD15; CD; CLR16.3; IBD1; NLRC2; NOD2; NOD2B; PSORAS1; YAOS

          View more View less

          UniProt ID: (Human) Q9HC29

          View more View less

          Entrez Gene ID: (Human) 64127

          View more View less

          Function(s)
          nucleotide binding actin binding protein binding ATP binding enzyme binding protein kinase binding Hsp70 protein binding muramyl dipeptide binding pattern recognition receptor activity peptidoglycan binding ubiquitin binding anion binding ADP binding macromolecular complex binding CARD domain binding Hsp90 protein binding carbohydrate derivative binding
          Process(es)
          temperature homeostasis pattern recognition receptor signaling pathway adaptive immune response immune system process positive regulation of dendritic cell antigen processing and presentation positive regulation of cytokine production involved in immune response positive regulation of dendritic cell cytokine production positive regulation of type 2 immune response autophagy defense response I-kappaB kinase/NF-kappaB signaling response to nutrient positive regulation of cell proliferation detection of biotic stimulus detection of bacterium maintenance of gastrointestinal epithelium regulation of appetite response to muramyl dipeptide detection of muramyl dipeptide positive regulation of interleukin-1 beta production positive regulation of interleukin-10 production positive regulation of interleukin-17 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production positive regulation of stress-activated MAPK cascade intracellular signal transduction intestinal stem cell homeostasis p38MAPK cascade defense response to bacterium regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of MAPK cascade innate immune response positive regulation of Notch signaling pathway positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade positive regulation of gamma-delta T cell activation host-mediated regulation of intestinal microbiota composition positive regulation of epithelial cell proliferation regulation of inflammatory response positive regulation of B cell activation positive regulation of NF-kappaB transcription factor activity positive regulation of ERK1 and ERK2 cascade nucleotide-binding oligomerization domain containing 2 signaling pathway protein K63-linked ubiquitination cellular response to lipopolysaccharide cellular response to peptidoglycan cellular response to muramyl dipeptide protein linear polyubiquitination antibacterial innate immune response positive regulation of cytokine production involved in inflammatory response positive regulation of NIK/NF-kappaB signaling positive regulation of mitophagy positive regulation of protein K63-linked ubiquitination negative regulation of macrophage apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline