Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NPC1L1 Antibodies

          Invitrogen

          NPC1L1 Polyclonal Antibody

          3 Published Figures
          4 References
          View all (6) NPC1L1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NPC1L1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (3)
          NPC1L1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NPC1L1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NPC1L1 Antibody (PA1-16800) in ICC/IF

          Immunocytochemistry analysis of NPC1L1 in HepG2 cells. Samples were incubated with NPC1L1 polyclonal antibody (Product # PA1-16800) using a dilution of 2 µg/mL overnight at 4°C followed by anti-rabbit Dylight 488 (Green) at a dilution of 1:500. Cells were fixed in 4% paraformaldehyde for 10 min and permeabilized in 0.05% Triton X-100 in PBS for 5 minutes. Nuclei were counterstained with DAPI (Blue). Cells were imaged using a 100X objective and digitally deconvolved. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NPC1L1 Antibody in Immunocytochemistry (ICC/IF)
          NPC1L1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NPC1L1 Antibody in Western Blot (WB)
          NPC1L1 Antibody in Immunocytochemistry (ICC/IF)
          NPC1L1 Antibody in Western Blot (WB)
          NPC1L1 Antibody in Western Blot (WB)
          NPC1L1 Polyclonal Antibody

          Product Details

          PA1-16800

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:250
          -

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 1000-1100).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7-8

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_568647

          Product Specific Information

          The target sequence has 93% sequence homology with human.

          Target Information

          Dietary consumption and intestinal cholesterol absorption contribute to plasma cholesterol levels, which factor into the risk of coronary heart disease. Niemann-Pick type C1 Like 1 protein (NPC1L1 or NPC3) is enriched in the small intestine and is found in the brush border membrane of enterocytes. It plays a critical role in absorption of intestinal cholesterol.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BC 400-127; cholesterol transporter; Niemann-Pick disease type C1; Niemann-Pick C1-like protein 1; Niemann-Pick C3; Niemann-Pick disease, type C1; Niemann-Pick disease, type C1-like 1; Niemann-Pick type C1-like 1 minus exon 15; NPC1 (Niemann-Pick disease, type C1, gene)-like 1; NPC1 like 1; NPC1-like intracellular cholesterol transporter 1; NPC1L1; NPC3; similar to Niemann-Pick C1 protein

          View more View less

          Gene Aliases: 9130221N23Rik; Gm243; LDLCQ7; NPC11L1; NPC1L1; SLC65A2

          View more View less

          UniProt ID: (Human) Q9UHC9, (Rat) Q6T3U3

          View more View less

          Entrez Gene ID: (Human) 29881, (Pig) 100515982, (Mouse) 237636, (Rat) 432367

          View more View less

          Function(s)
          protein binding vitamin E binding cholesterol binding small GTPase binding myosin V binding protein homodimerization activity drug binding hedgehog receptor activity Rab GTPase binding
          Process(es)
          lipid metabolic process cholesterol biosynthetic process steroid metabolic process cholesterol metabolic process intestinal cholesterol absorption cholesterol transport lipoprotein metabolic process cholesterol homeostasis sterol transport vitamin E metabolic process vitamin transport cellular response to sterol depletion signal transduction response to drug
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline