Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Npr2 Antibodies

          Invitrogen

          NPR2 Polyclonal Antibody

          1 Reference
          View all (13) Npr2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NPR2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          NPR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          NPR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NPR2 Antibody (PA5-99836) in IHC (P)

          Immunohistochemistry analysis of NPR2 in rat brain tissue. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with NPR2 polyclonal antibody (Product # PA5-99836) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NPR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NPR2 Antibody in Western Blot (WB)
          NPR2 Polyclonal Antibody

          Product Details

          PA5-99836

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human NPR2(Accession P20594), corresponding to amino acid residues S647-L697.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2818769

          Product Specific Information

          Antibody detects endogenous levels of total NPR2.

          Target Information

          This gene encodes natriuretic peptide receptor B, one of two integral membrane receptors for natriuretic peptides. Both NPR1 and NPR2 contain five functional domains: an extracellular ligand-binding domain, a single membrane-spanning region, and intracellularly a protein kinase homology domain, a helical hinge region involved in oligomerization, and a carboxyl-terminal guanylyl cyclase catalytic domain. The protein is the primary receptor for C-type natriuretic peptide, which upon ligand binding exhibits greatly increased guanylyl cyclase activity. Mutations in this gene are the cause of acromesomelic dysplasia Maroteaux type.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ANP-B; ANPR-B; ANPRB; atrial natriuretic peptide B-type receptor; Atrial natriuretic peptide receptor 2; atrial natriuretic peptide receptor B; Atrial natriuretic peptide receptor type B; GC-B; guanylate cyclase 2B; Guanylate cyclase B; guanylyl cyclase B; guanylyl cyclase-B; natriuretic peptide receptor B type; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); natriuretic peptide receptor-B; Nppb receptor; NPR-B

          View more View less

          Gene Aliases: AMD1; AMDM; ANPb; ANPRB; cn; ECDM; GC-B; GC-B2; GC-B3; GCB; GUC2B; GUCY2B; mNPR-B; NPR-B; NPR2; NPRB; NPRBi; pwe; SNSK

          View more View less

          UniProt ID: (Human) P20594, (Rat) P16067, (Mouse) Q6VVW5

          View more View less

          Entrez Gene ID: (Human) 4882, (Rat) 116564, (Mouse) 230103

          View more View less

          Function(s)
          nucleotide binding guanylate cyclase activity protein kinase activity protein binding ATP binding GTP binding lyase activity phosphorus-oxygen lyase activity natriuretic peptide receptor activity peptide hormone binding hormone binding identical protein binding transferase activity, transferring phosphorus-containing groups guanylate cyclase
          Process(es)
          MAPK cascade ossification ovarian follicle development cumulus cell differentiation blood vessel development vasculogenesis lymph vessel development endochondral ossification startle response blood vessel remodeling chondrocyte differentiation growth plate cartilage development cGMP biosynthetic process vacuole organization receptor guanylyl cyclase signaling pathway epidermal growth factor receptor signaling pathway chemical synaptic transmission germ cell development spermatogenesis single fertilization axonogenesis sensory perception of sound blood circulation regulation of blood pressure cyclic nucleotide biosynthetic process response to wounding response to hormone neuronal action potential vestibulocochlear nerve development vestibulocochlear nerve maturation nerve development female genitalia development response to luteinizing hormone limb morphogenesis genitalia morphogenesis multicellular organism growth gastric emptying intracellular signal transduction chondrocyte proliferation post-anal tail morphogenesis cGMP metabolic process digestive tract morphogenesis digestive tract development oocyte development collateral sprouting smooth muscle tissue development white fat cell differentiation cartilage development chromosome organization meiotic cell cycle neuron apoptotic process negative regulation of meiotic cell cycle limb development bone development axonogenesis involved in innervation activation of meiosis involved in egg activation vascular wound healing c-di-GMP signaling cellular response to cGMP response to fibroblast growth factor cellular response to granulocyte macrophage colony-stimulating factor stimulus execution phase of apoptosis bone growth negative regulation of oocyte maturation cellular response to peptide response to salt meiotic cell cycle process involved in oocyte maturation protein phosphorylation signal transduction single organism reproductive process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0