Recommended positive controls: NT2D1, IMR32, HeLa.
Predicted reactivity: Human (99%), Mouse (96%), Rat (95%), Zebrafish (80%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.