Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MTEYKLVVV GAGGVGKSAL TIQLIQNHFV DEYDPTIEDS YRKQVVIDGE TCLLDILDTA GQEEYSAMRD QYMRTGEGFL CVFAINNSKS FADINLYREQ IKRVKDSDDV PMVLVGNKCD LPTRTVDTKQ AHELAKSYGI PFIETSAKTR QGVEDAFYTL VREIRQYRMK KLNSSDDGTQ GCMGLPCVVM (1-189 aa encoded by BC005219)
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noonan syndrome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-ras protein part 4; Neuroblastoma RAS; neuroblastoma ras oncogene; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; Nras; proto-oncogene GTPase; RAS; RASN; small GTP binding protein; transforming protein N-Ras; unnamed protein product; v-ras neuroblastoma RAS viral oncogene homolog
Gene Aliases: ALPS4; CMNS; N-ras; NCMS; NRAS1; NS6; RALD1
UniProt ID: (Human) P01111, (Rat) Q04970
Entrez Gene ID: (Human) 4893, (Mouse) 18176, (Rat) 24605
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support