Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NUP62 Antibodies

          Proteintech

          NUP62 Polyclonal Antibody

          15 References
          View all (23) NUP62 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NUP62 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          NUP62 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NUP62 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NUP62 Antibody (13916-1-AP) in WB

          NIH/3T3 cells were subjected to SDS PAGE followed by western blot with 13916-1-AP (NUP62 antibody) at dilution of 1:1500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NUP62 Antibody in Western Blot (WB)
          NUP62 Antibody in Western Blot (WB)
          NUP62 Antibody in Immunoprecipitation (IP)
          NUP62 Polyclonal Antibody

          Product Details

          13916-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 9 publications 9 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NUP62 Fusion Protein Ag4912 (173-522 aa encoded by BC014842)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: IGSAGNSAQ PTAPATLPFT PATPAATTAG ATQPAAPTPT ATITSTGPSL FASIATAPTS SATTGLSLCT PVTTAGAPTA GTQGFSLKAP GAASGTSTTT STAATATATT TSSSSTTGFA LNLKPLAPAG IPSNTAAAVT APPGPGAAAG AAASSAMTYA QLESLINKWS LELEDQERHF LQQATQVNAW DRTLIENGEK ITSLHREVEK VKLDQKRLDQ ELDFILSQQK ELEDLLSPLE ELVKEQSGTI YLQHADEERE KTYKLAENID AQLKRMAQDL KDIIEHLNTS GAPADTSDPL QQICKILNAH MDSLQWIDQN SALLQRKVEE VTKVCEGRRK EQERSFRITF D (173-522 aa encoded by BC014842)

          Target Information

          SDS3 is a subunit of the histone deacetylase.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 62 kDa nucleoporin; DKFZp547L134; FLJ20822; FLJ43869; MGC841; nuclear pore complex 1; nuclear pore glycoprotein 62; Nuclear pore glycoprotein p62; nucleoporin 62kD; nucleoporin 62kDa; Nucleoporin Nup62; nucleoporin p62; NUP62; unnamed protein product

          View more View less

          Gene Aliases: AA589433; AI426861; AI790512; AU045898; D7Ertd649e; IBSN; Np62; NUP62; Nupc1; p62; SNDI

          View more View less

          UniProt ID: (Human) P37198, (Rat) P17955, (Mouse) Q63850

          View more View less

          Entrez Gene ID: (Human) 23636, (Rat) 65274, (Mouse) 18226

          View more View less

          Function(s)
          chromatin binding protein binding phospholipid binding structural constituent of nuclear pore receptor signaling complex scaffold activity Hsp70 protein binding SH2 domain binding ubiquitin binding PTB domain binding Hsp90 protein binding thyroid hormone receptor binding kinesin binding transporter
          Process(es)
          RNA export from nucleus protein import into nucleus nucleocytoplasmic transport mitotic metaphase plate congression centrosome cycle mitotic centrosome separation cell surface receptor signaling pathway negative regulation of cell proliferation regulation of signal transduction protein transport negative regulation of epidermal growth factor receptor signaling pathway negative regulation of apoptotic process negative regulation of programmed cell death positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of epidermal growth factor receptor signaling pathway positive regulation of mitotic nuclear division positive regulation of transcription, DNA-templated regulation of Ras protein signal transduction negative regulation of Ras protein signal transduction positive regulation of centriole replication mRNA transport regulation of mitotic spindle organization cellular senescence centriole assembly positive regulation of mitotic cytokinetic process positive regulation of protein localization to centrosome transcription, DNA-templated cell aging cell death negative regulation of MAP kinase activity transport cell migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-fcf5f9d6d-txc6f:80/100.66.78.195:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0