Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MA1-071 detects nuclear pore-O-linked glycoprotein from rat, amphibian and yeast.
MA1-071 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects up to eight different proteins from the nuclear pore complex (NPC) of approximately 210, 180, 145, 100, 63, 58, 54 and 45 kDa. Immunofluorescence staining of NPC O-linked glycoproteins with MA1-071 results in exclusive labeling of the NPC proteins on a wide variety of mammalian cells as well as S. cerevisiae and Xenopus. Labeling occurs exclusively at the NPC with most of the labeling at the cytoplasmic and nucleoplasmic margins.
Microinjected MA1-071 inhibits both protein import and RNA export in Xenopus oocytes.
The MA1-071 immunogen is pore complex-lamina fraction purified from rat liver nuclear envelopes.
SDS3 is a subunit of the histone deacetylase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 62 kDa nucleoporin; DKFZp547L134; FLJ20822; FLJ43869; MGC841; nuclear pore glycoprotein 62; nucleoporin Nup62; nucleoporin p62; nucleoporin p62 homolog
Gene Aliases: Np62
UniProt ID: (Rat) P17955
Entrez Gene ID: (Rat) 65274
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support