Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • NUP98 Antibodies

          Abnova

          NUP98 Polyclonal Antibody, MaxPab™

          View all (24) NUP98 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NUP98 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NUP98 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NUP98 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NUP98 Antibody (H00004928-D01P) in WB

          Western Blot analysis of NUP98 expression in transfected 293T cell line (H00004928-T02) by NUP98 MaxPab polyclonal antibody.

          Lane 1: NUP98 transfected lysate(69.80 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NUP98 Antibody in Western Blot (WB)
          NUP98 Polyclonal Antibody, MaxPab™

          Product Details

          H00004928-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NUP98 (AAH12906.1, 1 a.a. approximately 606 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MKLYQTPLEL KLKHSTVHVD ELCPLIVPNL GVAVIHDYAD WVKEASGDLP EAQIVKHWSL TWTLCEALWG HLKELDSQLN EPREYIQILE RRRAFSRWLS CTATPQIEEE VSLTQKNSPV EAVFSYLTGK RISEACSLAQ QSGDHRLALL LSQFVGSQSV RELLTMQLVD WHQLQADSFI QDERLRIFAL LAGKPVWQLS EKKQINVCSQ LDWKRSLAIH LWYLLPPTAS ISRALSMYEE AFQNTSDSDR YACSPLPSYL EGSGCVIAEE QNSQTPLRDV CFHLLKLYSD RHYDLNQLLE PRSITADPLD YRLSWHLWEV LRALNYTHLS AQCEGVLQAS YAGQLESEGL WEWAIFVLLH IDNSGIREKA VRELLTRHCQ LLETPESWAK ETFLTQKLRV PAKWIHEAKA VRAHMESDKH LEALCLFKAE HWNRCHKLII RHLASDAIIN ENYDYLKGFL EDLAPPERSS LIQDWETSGL VYLDYIRVIE MLRHIQQVDC SGNDLEQLHI KVTSLCSRIE QIQCYSAKDR LAQSDMAKRV ANLLRVVLSL HHPPDRTSDS TPDPQRVPLR LLAPHIGRLP MPEDYAMDEL RSLTQSYLRE LAVGSL

          Target Information

          Signal-mediated nuclear import and export proceed through the nuclear pore complex (NPC), which is comprised of approximately 50 unique proteins collectively known as nucleoporins. The 98 kDa nucleoporin is generated through a biogenesis pathway that involves synthesis and proteolytic cleavage of a 186 kDa precursor protein. This cleavage results in the 98 kDa nucleoporin as well as a 96 kDa nucleoporin, both of which are localized to the nucleoplasmic side of the NPC. Rat studies show that the 98 kDa nucleoporin functions as one of several docking site nucleoporins of transport substrates. The human gene has been shown to fuse to several genes following chromosome translocations in acute myelogenous leukemia (AML) and T-cell acute lymphocytic leukemia (T-ALL). This gene is one of several genes located in the imprinted gene domain of 11p15. 5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in several transcript variants; however, not all variants have been fully described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alternatively spliced; GLFG-repeat containing nucleoporin; Nuclear pore complex protein Nup98-Nup96; nucleoporin; nucleoporin 96; nucleoporin 98kD; nucleoporin 98kDa; Nup98-Nup96; NUP98/PHF23 fusion 2 protein; OTTHUMP00000013954; unnamed protein product

          View more View less

          Gene Aliases: ADAR2; ADIR2; NUP196; NUP96; NUP98; Nup98-96

          View more View less

          UniProt ID: (Human) P52948

          View more View less

          Entrez Gene ID: (Human) 4928

          View more View less

          Function(s)
          transcription coactivator activity RNA binding mRNA binding protein binding nuclear localization sequence binding peptidase activity serine-type peptidase activity hydrolase activity structural constituent of nuclear pore peptide binding molecular condensate scaffold activity promoter-specific chromatin binding
          Process(es)
          posttranscriptional tethering of RNA polymerase II gene DNA at nuclear periphery RNA export from nucleus proteolysis protein import into nucleus nucleocytoplasmic transport nuclear pore organization protein transport telomere tethering at nuclear periphery positive regulation of transcription, DNA-templated positive regulation of mRNA splicing, via spliceosome mRNA transport nuclear pore complex assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline