Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Nogo-D Antibodies

          Osenses

          Nogo Polyclonal Antibody

          2 References
          View all (5) Nogo-D antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Nogo Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          OSN00031W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-1:2,000
          -

          Immunohistochemistry (IHC)

          1:300-1:2,000
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Goat / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from internal region of human Nogo conjugated to an immunogenic carrier protein was used as the antigen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in rat and mouse.

          Specificity of this antibody: Nogo.

          Target Information

          Required to induce the formation and stabilization of endoplasmic reticulum (ER) tubules. They regulate membrane morphogenesis in the ER by promoting tubular ER production. They influence nuclear envelope expansion, nuclear pore complex formation and proper localization of inner nuclear membrane proteins. However each isoform have specific functions mainly depending on their tissue expression specificities. Isoform D is expressed at very low levels in neurons.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ASY; Foocen; Glut4 vesicle 20 kDa protein; GLUT4 vesicle 20kDa protein; Human NogoA; My043 protein; neurite growth inhibitor 220; Neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; NI-220/250; Nogo protein; NOGO-A; RTN4-A; NOGO-B; RTN4-B1; NOGO-C; RTN4-C; NSP; reticulon; reticulon 5; Reticulon-4; Reticulon-5; RTN-x; RTN4-Aa; RTN4-Ab; RTN4-B2; RTN4-D; RTN4-E; RTN4-F; RTN4-G; RTN4a; RTN4b; RTN4c; unnamed protein product

          View more View less

          Gene Aliases: 1110020G17Rik; AA407876; AA409940; AA960376; ASY; C130026I10Rik; KIAA0886; mKIAA0886; mKIAA4153; My043; Nbla00271; Nbla10545; NgA; NI-250; NI220/250; NOGO; Nogo-A; Nogo-B; Nogo-C; NSP; NSP-CL; r; rat N; rat NogoA; RTN-X; RTN4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; SP1507; Vp20

          View more View less

          UniProt ID: (Human) Q9NQC3, (Mouse) Q99P72, (Rat) Q9JK11

          View more View less

          Entrez Gene ID: (Human) 57142, (Mouse) 68585, (Rat) 83765

          View more View less

          Function(s)
          RNA binding protein binding ubiquitin protein ligase binding protein homodimerization activity cadherin binding metal ion binding protein complex binding poly(A) RNA binding
          Process(es)
          angiogenesis nervous system development axonogenesis axonal fasciculation negative regulation of neuron projection development cerebral cortex radial glia guided migration negative regulation of cell growth regulation of cell migration negative regulation of axon extension positive regulation of dopamine secretion negative regulation of neuron differentiation positive regulation of glial cell differentiation negative regulation of axonogenesis regulation of sensory perception of pain regulation of nervous system development cardiac epithelial to mesenchymal transition negative regulation of neuron projection regeneration endoplasmic reticulum tubular network organization regulation of branching morphogenesis of a nerve blastocyst formation leukocyte migration involved in inflammatory response apoptotic process endoplasmic reticulum organization brain development positive regulation of epithelial cell migration central nervous system vasculogenesis neuron differentiation positive regulation of mammary gland epithelial cell proliferation positive regulation of toll-like receptor 9 signaling pathway positive regulation of Rac protein signal transduction cell migration involved in vasculogenesis regulation of apoptotic process positive regulation of angiogenesis modulation of synaptic transmission protein stabilization nuclear pore complex assembly positive regulation of protein kinase B signaling positive regulation of macrophage cytokine production protein localization to lysosome cellular response to hypoxia endoplasmic reticulum tubular network assembly cellular sphingolipid homeostasis cell adhesion involved in sprouting angiogenesis regulation of postsynapse assembly negative regulation of beta-amyloid formation positive regulation of neutrophil migration positive regulation of macrophage migration positive regulation of protein localization to endoplasmic reticulum positive regulation of ERBB3 signaling pathway positive regulation of artery morphogenesis endoplasmic reticulum tubular network membrane organization positive regulation of hepatocyte proliferation negative regulation of vasculogenesis aging response to activity olfactory nerve development oligodendrocyte differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline