Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: QQLKASVKN YEGMIDNYKS QVMKTRLEAD EVAAQLERCD KENKILKDEM NKEIEAARRQ FQSQLADLQQ LPDILKITEA KLAECQDQLQ GYERKNIDLT AIISDLRSRI EHQGDKLEMA REKHQASQKE NKQLSLKVDE LERKLEATSA QNIEFLQVIA KREEAIHQSQ LRLEEKTREC GTLARQLESA IEDARRQVEQ TKEHALSKER AAQNKILDLE TQLSRTKTEL SQLRRSRDDA DRRYQSRLQD LKDRLEQSES TNRSMQNYVQ FLKSSYANVF GDGPYSTFLT SSPIRSRSPP A (530-829 aa encoded by BC010629)
Seems to be a major component of sperm tail outer dense fibers (ODF). ODFs are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail and may help to maintain the passive elastic structures and elastic recoil of the sperm tail. May have a modulating influence on sperm motility. Functions as a general scaffold protein that is specifically localized at the distal/subdistal appendages of mother centrioles. Component of the centrosome matrix required for the localization of PLK1 and NIN to the centrosomes. Required for the formation and/or maintenance of normal CETN1 assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 84 kDa outer dense fiber protein; cancer/testis antigen 134; cenexin; cenexin 1; hCenexin; ODF2; Odf2/2; ORF; outer dense fiber of sperm tail 2; outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; outer dense filament; RP11-187B3.1; sperm outer dense fiber major protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e; testis-specific autoantigen; unnamed protein product
Gene Aliases: CT134; MMTEST29; ODF2/1; ODF2/2; ODF84
UniProt ID: (Human) Q5BJF6, (Mouse) A3KGV1, (Rat) Q6AYX5
Entrez Gene ID: (Human) 4957, (Mouse) 18286, (Rat) 29479
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support