Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • OGT Antibodies

          Proteintech

          OGT Polyclonal Antibody

          53 References
          View all (17) OGT antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite OGT Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (20)
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OGT Antibody (11576-2-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human colon cancer tissue slide using (Product # 11576-2-AP) (OGT antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OGT Antibody in Western Blot (WB)
          OGT Antibody in Western Blot (WB)
          OGT Antibody in Western Blot (WB)
          OGT Antibody in Western Blot (WB)
          OGT Antibody in Western Blot (WB)
          OGT Antibody in Western Blot (WB)
          OGT Antibody in Western Blot (WB)
          OGT Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          OGT Polyclonal Antibody

          Product Details

          11576-2-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 31 publications 31 publications

          Immunohistochemistry (IHC)

          -
          View 10 publications 10 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 7 publications 7 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          OGT Fusion Protein Ag2160 (1-400 aa encoded by BC014434)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.37 mg/mL

          Amount

          55.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MASSVGNVA DSTGLAELAH REYQAGDFEA AERHCMQLWR QEPDNTGVLL LLSSIHFQCR RLDRSAHFST LAIKQNPLLA EAYSNLGNVY KERGQLQEAI EHYRHALRLK PDFIDGYINL AAALVAAGDM EGAVQAYVSA LQYNPDLYCV RSDLGNLLKA LGRLEEAKAC YLKAIETQPN FAVAWSNLGC VFNAQGEIWL AIHHFEKAVT LDPNFLDAYI NLGNVLKEAR IFDRAVAAYL RALSLSPNHA VVHGNLACVY YEQGLIDLAI DTYRRAIELQ PHFPDAYCNL ANALKEKGSV AEAEDCYNTA LRLCPTHADS LNNLANIKRE QGNIEEAVRL YRKALEVFPE FAAAHSNLAS VLQQQGKLQE ALMHYKEAIR ISPTFADAYS NMGNTLKEMQ D (1-400 aa encoded by BC014434)

          Target Information

          This gene encodes a glycosyltransferase that catalyzes the addition of a single N-acetylglucosamine in O-glycosidic linkage to serine or threonine residues. Since both phosphorylation and glycosylation compete for similar serine or threonine residues, the two processes may compete for sites, or they may alter the substrate specificity of nearby sites by steric or electrostatic effects. The protein contains multiple tetratricopeptide repeats that are required for optimal recognition of substrates. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FLJ23071; MGC22921; O linked N-acetylglucosamine transferase; o-glcnac transferase; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; OGT; O-GlcNAc transferase; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; unnamed protein product; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase

          View more View less

          Gene Aliases: 1110038P24Rik; 4831420N21Rik; HINCUT-1; HRNT1; MRX106; O-GLCNAC; OGT1; Ogtl; XLID106

          View more View less

          UniProt ID: (Human) O15294, (Mouse) Q8CGY8, (Rat) P56558

          View more View less

          Entrez Gene ID: (Human) 8473, (Mouse) 108155, (Rat) 26295

          View more View less

          Function(s)
          protein binding phosphatidylinositol-3,4,5-trisphosphate binding acetylglucosaminyltransferase activity transferase activity, transferring glycosyl groups chromatin DNA binding protein O-GlcNAc transferase activity catalytic activity N-acetyltransferase activity protein domain specific binding peptide binding identical protein binding macromolecular complex binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter mitophagy positive regulation of transcription from RNA polymerase II promoter by glucose regulation of glycolytic process regulation of gluconeogenesis regulation of transcription from RNA polymerase II promoter protein O-linked glycosylation apoptotic process signal transduction response to nutrient protein processing hemopoiesis negative regulation of cell migration negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of protein ubiquitination negative regulation of proteasomal ubiquitin-dependent protein catabolic process response to insulin circadian regulation of gene expression regulation of Rac protein signal transduction positive regulation of translation positive regulation of proteolysis positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter regulation of insulin receptor signaling pathway regulation of synapse assembly regulation of necroptotic process cellular response to glucose stimulus regulation of neurotransmitter receptor localization to postsynaptic specialization membrane positive regulation of cold-induced thermogenesis negative regulation of non-canonical inflammasome complex assembly negative regulation of stem cell population maintenance positive regulation of stem cell population maintenance positive regulation of TORC1 signaling negative regulation of protein phosphorylation positive regulation of protein phosphorylation cytoplasmic translation glucosamine metabolic process positive regulation of gene expression negative regulation of peptidyl-threonine phosphorylation negative regulation of translation cellular response to nutrient levels negative regulation of peptidyl-serine phosphorylation TORC1 signaling positive regulation of cell size negative regulation of translational initiation positive regulation of translational initiation intracellular distribution of mitochondria protein maturation regulation of cellular localization protein localization to lysosome pyroptosis cellular response to lipopolysaccharide negative regulation of protein targeting to membrane membraneless organelle assembly non-canonical inflammasome complex assembly negative regulation of cellular response to hypoxia positive regulation of protein localization to nucleus positive regulation of reactive oxygen species biosynthetic process forebrain development cellular response to insulin stimulus cellular response to toxic substance
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-684z6:80/100.66.78.224:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0