Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Application for antibody includes Stable Isotope Standards and Capture by Anti-Peptide Antibodies (SISCAPA).
OLFM4 is a 510 amino acid extracellular matrix glycoprotein belonging to the olfactomedin-related protein family and found to be selectively expressed in inflammed colonic epithelium. It has one signal peptide, six N-linked glycosylation motif and forms disulfide-bonded multimers. OLFM4 a transcription factor may have a matrix-related function in differentiation. Constitutively expressed OLFM4 in cancer cells binds to the potent apoptosis inducer GRIM-19 and has a novel antiapoptotic action and hence might serve as a diagnostic marker for human cancers. Reports suggest that OLFM4 promotes proliferation of PANC-1 cells by favoring transition from the S to G2/M phase. Potential interaction of OLFM4 with endogenous cell surface lectins and cadherins may mediate cell adhesion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antiapoptotic protein GW112; G-CSF-stimulated clone 1 protein; hGC-1; hOLfD; Olfactomedin-4; OLM4
Gene Aliases: bA209J19.1; GC1; GW112; hGC-1; hOLfD; OlfD; OLFM4; OLM4; UNQ362; UNQ362/PRO698
UniProt ID: (Human) Q6UX06
Entrez Gene ID: (Human) 10562
Molecular Function: structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support