Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • OPA1 Antibodies

          Abnova

          OPA1 Polyclonal Antibody, MaxPab™

          View all (15) OPA1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite OPA1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          OPA1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          OPA1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OPA1 Antibody (H00004976-D01P) in WB

          Western Blot analysis of OPA1 expression in transfected 293T cell line by OPA1 MaxPab polyclonal antibody. Lane 1: OPA1 transfected lysate(111.60 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OPA1 Antibody in Western Blot (WB)
          OPA1 Polyclonal Antibody, MaxPab™

          Product Details

          H00004976-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          OPA1 (NP_056375.1, 1 a.a. approximately 960 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MWRLRRAAVA CEVCQSLVKH SSGIKGSLPL QKLHLVSRSI YHSHHPTLKL QRPQLRTSFQ QFSSLTNLPL RKLKFSPIKY GYQPRRNFWP ARLATRLLKL RYLILGSAVG GGYTAKKTFD QWKDMIPDLS EYKWIVPDIV WEIDEYIDFE KIRKALPSSE DLVKLAPDFD KIVESLSLLK DFFTSGSPEE TAFRATDRGS ESDKHFRKVS DKEKIDQLQE ELLHTQLKYQ RILERLEKEN KELRKLVLQK DDKGIHHRKL KKSLIDMYSE VLDVLSDYDA SYNTQDHLPR VVVVGDQSAG KTSVLEMIAQ ARIFPRGSGE MMTRSPVKVT LSEGPHHVAL FKDSSREFDL TKEEDLAALR HEIELRMRKN VKEGCTVSPE TISLNVKGPG LQRMVLVDLP GVINTVTSGM APDTKETIFS ISKAYMQNPN AIILCIQDGS VDAERSIVTD LVSQMDPHGR RTIFVLTKVD LAEKNVASPS RIQQIIEGKL FPMKALGYFA VVTGKGNSSE SIEAIREYEE EFFQNSKLLK TSMLKAHQVT TRNLSLAVSD CFWKMVRESV EQQADSFKAT RFNLETEWKN NYPRLRELDR NELFEKAKNE ILDEVISLSQ VTPKHWEEIL QQSLWERVST HVIENIYLPA AQTMNSGTFN TTVDIKLKQW TDKQLPNKAV EVAWETLQEE FSRFMTEPKG KEHDDIFDKL KEAVKEESIK RHKWNDFAED SLRVIQHNAL EDRSISDKQQ WDAAIYFMEE ALQARLKDTE NAIENMVGPD WKKRWLYWKN RTQEQCVHNE TKNELEKMLK CNEEHPAYLA SDEITTVRKN LESRGVEVDP SLIKDTWHQV YRRHFLKTAL NHCNLCRRGF YYYQRHFVDS ELECNDVVLF WRIQRMLAIT ANTLRQQLTN TEVRRLEKNV KEVLEDFAED GEKKIKLLTG KRVQLAEDLK KVREIQEKLD AFIEALHQEK

          Target Information

          OPA1 is a dynamin-related GTPase that is critical for the maintenance of mitochondrial morphology and mtDNA. The most commonly associated phenotype with OPA1 mutations is heterozygous optic atrophy, a heterozygous dominant trait that causes reduced visual clarity and sometimes blindness. The disease usually begins in childhood and increases in severity throughout the life of affected individuals. Usually, this phenotype is attributed to the degeneration of optic nerve fibers. Interestingly, the same type of nerve degeneration seems to be partially causative of certain schizophrenia characteristics. OPA1 dysfunction also seems to be implicated in this case; mitochondrial networks associated with critical nerves seem to link schizophrenia and OPA1. The dysfunction is associated with issues with apoptosis and normal cellular metabolic regulation, all regulated through OPA1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: dynamin-like 120 kDa protein, mitochondrial; Dynamin-like GTPase OPA1, mitochondrial; dynamin-like guanosine triphosphatase; mitochondrial dynamin-like GTPase; optic atrophy 1 (autosomal dominant); Optic atrophy protein 1

          View more View less

          Gene Aliases: BERHS; KIAA0567; largeG; MGM1; MTDPS14; NPG; NTG; OPA1

          View more View less

          UniProt ID: (Human) O60313

          View more View less

          Entrez Gene ID: (Human) 4976

          View more View less

          Function(s)
          nucleotide binding magnesium ion binding GTPase activity protein binding GTP binding microtubule binding lipid binding hydrolase activity phosphatidic acid binding GTPase-dependent fusogenic activity membrane bending activity cardiolipin binding
          Process(es)
          mitochondrial genome maintenance mitochondrial fission neural tube closure apoptotic process mitochondrion organization inner mitochondrial membrane organization visual perception mitochondrial fusion regulation of mitochondrion organization peroxisome fission axonal transport of mitochondrion positive regulation of interleukin-17 production cristae formation negative regulation of apoptotic process GTP metabolic process protein oligomerization membrane fusion negative regulation of release of cytochrome c from mitochondria cellular senescence membrane tubulation negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway mitochondrial inner membrane fusion positive regulation of T-helper 17 cell lineage commitment negative regulation of intrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline