Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • P2X7 Antibodies

          Huabio

          P2X7 Polyclonal Antibody

          View all (26) P2X7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite P2X7 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          P2X7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          P2X7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          P2X7 Antibody (ER1901-99) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human kidney tissue using anti-P2X7 antibody. The section was pre-treated using heat mediated antigen retrieval with Tris-EDTA buffer (pH 8.0-8.4) for 20 minutes. The tissues were blocked in 5% BSA for 30 minutes at room temperature, washed with ddH2O and PBS, and then probed with the primary antibody (ER1901-99, 1:50) for 30 minutes at room temperature. The... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          P2X7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P2X7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P2X7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P2X7 Antibody in Western Blot (WB)
          P2X7 Antibody in Flow Cytometry (Flow)
          P2X7 Polyclonal Antibody

          Product Details

          ER1901-99

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to C terminal Rat P2X7.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 0.2% BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Multiple alternatively spliced variants have been identified, most of which fit nonsense-mediated decay criteria.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATP agonist non-selective cationic pore-forming protein; ATP receptor; ligand gated ion channel; MGC20089; P2X purinoceptor 7; p2X receptor 7; P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7; cervical cancer susceptibility protein; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X7; purnergic receptor P2X 7; unnamed protein product

          View more View less

          Gene Aliases: AI467586; P2RX7; P2X(7); P2X7; P2X7R

          View more View less

          UniProt ID: (Human) Q99572, (Rat) Q64663, (Mouse) Q9Z1M0

          View more View less

          Entrez Gene ID: (Human) 5027, (Rat) 29665, (Mouse) 18439

          View more View less

          Function(s)
          nucleotide binding lipopolysaccharide binding purinergic nucleotide receptor activity extracellular ATP-gated cation channel activity receptor binding ion channel activity potassium channel activity sodium channel activity protein binding ATP binding GTP binding channel activity signaling receptor activity identical protein binding metal ion binding magnesium ion binding receptor activity copper ion binding zinc ion binding protein serine/threonine kinase activator activity scaffold protein binding ligand-gated ion channel
          Process(es)
          MAPK cascade cell morphogenesis phagolysosome assembly T cell mediated cytotoxicity positive regulation of T cell mediated cytotoxicity regulation of sodium ion transport response to ischemia membrane protein ectodomain proteolysis phospholipid transfer to membrane ion transport cation transport calcium ion transport cell volume homeostasis membrane budding inflammatory response mitochondrion organization plasma membrane organization cell surface receptor signaling pathway protein secretion response to xenobiotic stimulus response to mechanical stimulus response to bacterium response to zinc ion gene expression positive regulation of calcium ion transport into cytosol positive regulation of gene expression programmed cell death glutamate secretion positive regulation of glutamate secretion gamma-aminobutyric acid secretion positive regulation of gamma-aminobutyric acid secretion organophosphate ester transport synaptic vesicle exocytosis protein processing phospholipid scrambling sensory perception of pain calcium-mediated signaling protein catabolic process positive regulation of bone mineralization bleb assembly positive regulation of prostaglandin secretion prostaglandin secretion response to lipopolysaccharide positive regulation of interleukin-1 alpha production positive regulation of interleukin-1 beta production positive regulation of interleukin-6 production collagen metabolic process response to ATP ion transmembrane transport response to fluid shear stress positive regulation of ion transmembrane transport purinergic nucleotide receptor signaling pathway sodium ion transmembrane transport T cell proliferation T cell homeostasis positive regulation of apoptotic process NAD transport negative regulation of MAPK cascade positive regulation of MAPK cascade phospholipid translocation positive regulation of ossification negative regulation of bone resorption negative regulation of cell volume positive regulation of glycolytic process ceramide biosynthetic process pore complex assembly skeletal system morphogenesis homeostasis of number of cells within a tissue positive regulation of protein secretion defense response to Gram-positive bacterium regulation of transport positive regulation of cellular component organization release of sequestered calcium ion into cytosol positive regulation of cytoskeleton organization response to calcium ion response to electrical stimulus establishment of localization in cell mitochondrial depolarization membrane depolarization positive regulation of mitochondrial depolarization excitatory postsynaptic potential positive regulation of macrophage cytokine production protein homotrimerization lymphocyte apoptotic process positive regulation of lymphocyte apoptotic process T cell apoptotic process positive regulation of T cell apoptotic process calcium ion transmembrane transport cellular response to ATP cellular response to dsRNA potassium ion transmembrane transport reactive oxygen species metabolic process apoptotic signaling pathway extrinsic apoptotic signaling pathway cation transmembrane transport positive regulation of NLRP3 inflammasome complex assembly positive regulation of bleb assembly activation of MAPK activity positive regulation of protein phosphorylation protein phosphorylation response to organic substance response to organic cyclic compound organic cation transport neuronal action potential cytolysis cellular response to extracellular stimulus response to drug positive regulation of catalytic activity multicellular organismal protein catabolic process positive regulation of cytolysis positive regulation of cytokine secretion positive regulation of interleukin-1 alpha secretion positive regulation of interleukin-1 beta secretion protein oligomerization cellular response to organic cyclic compound transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0