Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • P2Y12 Antibodies

          Alomone Labs, LTD

          P2Y12 Receptor (extracellular) Polyclonal Antibody, APC

          View all (33) P2Y12 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite P2Y12 Receptor (extracellular) Polyclonal Antibody, APC

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          P2Y12 Receptor (extracellular) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          P2Y12 Receptor (extracellular) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          P2Y12 Receptor (extracellular) Antibody (APR-020-APC-50UL) in Flow

          Cell surface multicolor flow cytometry analysis of live intact mouse BV-2 microglia cells with P2Y12 and TREM2. BV-2 cells were labeled with Anti-P2Y12 Receptor (extracellular)-APC Antibody (#APR-020-APC) (5 µg) and with Anti-TREM2 (extracellular)-PE Antibody (#ANR-018-PE) (5 µg). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          P2Y12 Receptor (extracellular) Antibody in Flow Cytometry (Flow)
          P2Y12 Receptor (extracellular) Antibody in Flow Cytometry (Flow)
          P2Y12 Receptor (extracellular) Antibody in Flow Cytometry (Flow)
          P2Y12 Receptor (extracellular) Polyclonal Antibody, APC

          Product Details

          APR-020-APC-50UL

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          2.5-5 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide EFRQEEFRKLAGR, corresponding to amino acid residues 2-14 of rat VGLUT1. Intracellular, N-terminus.
          3D Epitope / Immunogen

          Conjugate

          APC APC APC

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Lyophilized

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Immunogen sequence: EFRQEEFRKLAGR

          Reconstitution: 25 uL, 50 uL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          P2Y12 belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is involved in platelets aggregation, and is a potential target for the treatment of thromboembolisms and other clotting disorders. Two transcript variants encoding the same isoform have been identified for this gene. Expression of P2Y12 has been reported in platelets, spinal cord, and many brain regions. ESTs for P2Y12 have been isolated from B-cell/lung/testis, brain, embryo, prostate, eye, kidney carcinoma, and colon carcinoma libraries.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ADP-glucose receptor; ADPG-R; G protein-coupled receptor; G-protein coupled receptor SP1999; Gi-coupled ADP receptor HORK3; P2T(AC); P2Y purinoceptor 12; P2Y(AC); P2Y(ADP); P2Y(cyc); P2Y12; P2Y12 platelet ADP receptor; P2YR12; purinergic receptor P2RY12; purinergic receptor P2Y, G-protein coupled, 12; putative G-protein coupled receptor; SP1999; unnamed protein product

          View more View less

          Gene Aliases: 2900079B22Rik; 4921504D23Rik; ADPG-R; BDPLT8; HORK3; P2RY12; P2T(AC); P2Y(12)R; P2Y(AC); P2Y(ADP); P2Y(cyc); P2Y12; SP1999

          View more View less

          UniProt ID: (Human) Q9H244, (Mouse) Q9CPV9, (Rat) Q9EPX4

          View more View less

          Entrez Gene ID: (Human) 64805, (Mouse) 70839, (Rat) 64803

          View more View less

          Function(s)
          G-protein coupled adenosine receptor activity ADP receptor activity G-protein coupled receptor activity guanyl-nucleotide exchange factor activity G-protein coupled purinergic nucleotide receptor activity signal transducer activity G-protein coupled receptor
          Process(es)
          adenosine receptor signaling pathway ion transport substrate-dependent cell migration, cell extension signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway blood coagulation hemostasis calcium-mediated signaling cerebral cortex radial glia guided migration cell projection organization lamellipodium assembly platelet activation positive regulation of integrin activation by cell surface receptor linked signal transduction positive regulation of cell adhesion mediated by integrin G-protein coupled purinergic nucleotide receptor signaling pathway positive regulation of ion transport response to axon injury regulation of chemotaxis positive regulation of chemotaxis establishment of localization in cell positive regulation of protein kinase B signaling platelet aggregation cellular response to ATP visual system development positive regulation of ruffle assembly regulation of microglial cell migration positive regulation of microglial cell migration glial cell migration negative regulation of norepinephrine secretion protein kinase B signaling negative regulation of cell differentiation regulation of calcium ion transport calcium ion transmembrane transport cellular response to organic cyclic compound potassium ion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline