Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: PEHQRANGN LKYFEYIMAK EKDVNKSASD DQSDQKTTPK KKGVAVDYLP ERQKYEMLCR GEGIKMTPRR QKKLFCRYHD GNRNPKFILA PAKQEDEWDK PRIIRFHDII SDAEIEIVKD LAKPRLRRAT ISNPITGDLE TVHYRISKSA WLSGYENPVV SRINMRIQDL TGLDVSTAEE LQVANYGVGG QYEPHFDFAR KDEPDAFKEL GTGNRIATWL FYMSDVSAGG ATVFPEVGAS VWPKKGTAVF WYNLFASGEG DYSTRHAACP VLVGNKWVSN KWLHERGQEF RRPCTLSELE (236-534 aa encoded by BC034998)
This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4-PH alpha-1; C-P4Halpha(I); collagen prolyl 4-hydroxylase alpha(I); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha 1 polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide I; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-1; prolyl 4-hydroxylase alpha subunit; prolyl 4-hydroxylase alpha subunit (EC 1.14.11.2); prolyl 4-hydroxylase, alpha polypeptide I; unnamed protein product
Gene Aliases: P4HA; PHalphaI
UniProt ID: (Human) P13674, (Rat) P54001, (Mouse) Q60715
Entrez Gene ID: (Human) 5033, (Rat) 64475, (Mouse) 18451
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support