Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • P4HTM Antibodies

          Abnova

          P4HTM Polyclonal Antibody, MaxPab™

          View all (12) P4HTM antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite P4HTM Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          P4HTM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          P4HTM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          P4HTM Antibody (H00054681-B01P) in WB

          Western Blot analysis of P4HTM expression in transfected 293T cell line by P4HTM MaxPab polyclonal antibody. Lane 1: P4HTM transfected lysate(61.93 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          P4HTM Antibody in Western Blot (WB)
          P4HTM Polyclonal Antibody, MaxPab™

          Product Details

          H00054681-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          P4HTM (NP_808807.2, 1 a.a. approximately 563 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAAAVTGQR PETAAAEEAS RPQWAPPDHC QAQAAAGLGD GEDAPVRPLC KPRGICSRAY FLVLMVFVHL YLGNVLALLL FVHYSNGDES SDPGPQHRAQ GPGPEPTLGP LTRLEGIKVG HERKVQLVTD RDHFIRTLSL KPLLFEIPGF LTDEECRLII HLAQMKGLQR SQILPTEEYE EAMSTMQVSQ LDLFRLLDQN RDGHLQLREV LAQTRLGNGW WMTPESIQEM YAAIKADPDG DGVLSLQEFS NMDLRDFHKY MRSHKAESSE LVRNSHHTWL YQGEGAHHIM RAIRQRVLRL TRLSPEIVEL SEPLQVVRYG EGGHYHAHVD SGPVYPETIC SHTKLVANES VPFETSCRQV SPNWGLPSIL RPGTPMTQAQ PCTVGVPLGM GPGDHWVIPV SPWEHPQLGT CSVPPLPYSY MTVLFYLNNV TGGGETVFPV ADNRTYDEMS LIQDDVDLRD TRRHCDKGNL RVKPQQGTAV FWYNYLPDGQ GWVGDVDDYS LHGGCLVTRG TKWIANNWIN VDPSRARQAL FQQEMARLAR EGGTDSQPEW ALDRAYRDAR VEL

          Target Information

          Prolyl hydroxylase 4 is a prolyl hydroxylase that modifies HIF-alpha. Classic prolyl hydroxylases are found in the endoplasmic reticulum and modify collagen, whereas HIF is an intracellular protein and the prolyl hydroxylase sites do not resemble those modifying collagen. HIF is a transcriptional complex that plays a critical role in oxygen homeostasis. Prolyl hydroxylase is an essential component of the pathway through which cells sense oxygen. In the presence of oxygen, prolyl hydroxylases convert specific prolyl residues in HIF-alpha to hydroxyproline, leading to HIF-alpha destruction. Low oxygen levels, sensed at the cellular level, cause the HIF conversion to be reduced so that HIF is stable and there is increased angiogenesis. Prolyl hydroxylase 4, specifically, catalyzes the posttranslational formation of 4-hydroxyproline in HIF alpha proteins. It may function as a cellular oxygen sensor and, under normoxic conditions, may targets HIF through the hydroxylation for proteasomal degradation via the von Hippel-Lindau ubiquitylation complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ20262; HIF-PH4; HIF-prolyl hydroxylase 4; HPH-4; hypoxia-inducible factor prolyl 4-hydroxylase; Hypoxia-inducible factor prolyl hydroxylase 4; P4H TM; P4H with transmembrane domain; P4H-TM; PH 4; proline 4-hydroxylase; prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum); Prolyl hydroxlase domain-containing 4; Transmembrane prolyl 4-hydroxylase; unnamed protein product

          View more View less

          Gene Aliases: EGLN4; HIDEA; HIFPH4; P4H-TM; P4HTM; PH-4; PH4; PHD4

          View more View less

          UniProt ID: (Human) Q9NXG6

          View more View less

          Entrez Gene ID: (Human) 54681

          View more View less

          Function(s)
          procollagen-proline 4-dioxygenase activity iron ion binding calcium ion binding oxidoreductase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors L-ascorbic acid binding metal ion binding dioxygenase activity hypoxia-inducible factor-proline dioxygenase activity
          Process(es)
          regulation of erythrocyte differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline