Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • SQSTM1 Antibodies

          Proteintech

          P62/SQSTM1 Polyclonal Antibody

          1,353 References
          View all (51) SQSTM1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite P62/SQSTM1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (28)
          P62/SQSTM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          P62/SQSTM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 28

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          P62/SQSTM1 Antibody (18420-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed Chloroquine treated HeLa cells using P62,SQSTM1 antibody (Product # 18420-1-AP) at dilution of 1:500 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          P62/SQSTM1 Antibody in Immunocytochemistry (ICC/IF)
          P62/SQSTM1 Antibody in Immunocytochemistry (ICC/IF)
          P62/SQSTM1 Antibody in Immunocytochemistry (ICC/IF)
          P62/SQSTM1 Antibody in Immunocytochemistry (ICC/IF)
          P62/SQSTM1 Antibody in Immunocytochemistry (ICC/IF)
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          P62/SQSTM1 Antibody in Western Blot (WB)
          P62/SQSTM1 Antibody in Western Blot (WB)
          P62/SQSTM1 Antibody in Western Blot (WB)
          P62/SQSTM1 Antibody in Western Blot (WB)
          P62/SQSTM1 Antibody in Western Blot (WB)
          P62/SQSTM1 Antibody in Western Blot (WB)
          P62/SQSTM1 Antibody in Western Blot (WB)
          P62/SQSTM1 Antibody in Flow Cytometry (Flow)
          P62/SQSTM1 Antibody in Immunoprecipitation (IP)
          P62/SQSTM1 Polyclonal Antibody

          Product Details

          18420-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 1,062 publications 1,062 publications

          Immunohistochemistry (IHC)

          -
          View 118 publications 118 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 134 publications 134 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 19 publications 19 publications

          Miscellaneous PubMed (Misc)

          -
          View 6 publications 6 publications

          Not applicable (N/A)

          -
          View 14 publications 14 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Chicken, Human, Mouse, Pig, Rabbit, Rat, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          P62/SQSTM1 Fusion Protein Ag13131 (1-440 aa encoded by BC017222)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.29 mg/mL

          Amount

          43.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          The molecular weight of p62 is predicted to be 48/ 38 kDa (depending on the isoform), while western blot analyses using this antibody detects the bands around 45-48 kDa and 60-62 kDa, respectively.

          Immunogen sequence: MASLTVKAY LLGKEDAARE IRRFSFCCSP EPEAEAEAAA GPGPCERLLS RVAALFPALR PGGFQAHYRD EDGDLVAFSS DEELTMAMSY VKDDIFRIYI KEKKECRRDH RPPCAQEAPR NMVHPNVICD GCNGPVVGTR YKCSVCPDYD LCSVCEGKGL HRGHTKLAFP SPFGHLSEGF SHSRWLRKVK HGHFGWPGWE MGPPGNWSPR PPRAGEARPG PTAESASGPS EDPSVNFLKN VGESVAAALS PLGIEVDIDV EHGGKRSRLT PVSPESSSTE EKSSSQPSSC CSDPSKPGGN VEGATQSLAE QMRKIALESE GRPEEQMESD NCSGGDDDWT HLSSKEVDPS TGELQSLQMP ESEGPSSLDP SQEGPTGLKE AALYPHLPPE ADPRLIESLS QMLSMGFSDE GGWLTRLLQT KNYDIGAALD TIQYSKHPPP L (1-440 aa encoded by BC017222)

          Target Information

          This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: autophagy receptor p62; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; oxidative stress induced; oxidative stress induced like; p62; p62 transcript variant 4; p62 transcript variant 5; phosphotyrosine independent ligand for the Lck SH2 domain p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; Protein kinase C-zeta-interacting protein; Sequestosome-1; STONE14; Ubiquitin-binding protein p62; unnamed protein product

          View more View less

          Gene Aliases: A170; DMRV; EBIAP; FTDALS3; NADGP; ORCA; OSF-6; Osi; OSIL; p60; p62; p62B; PDB3; SQSTM1; STAP; STONE14; ZIP; ZIP3

          View more View less

          UniProt ID: (Human) Q13501, (Rat) O08623, (Mouse) Q64337

          View more View less

          Entrez Gene ID: (Human) 8878, (Rat) 113894, (Mouse) 18412

          View more View less

          Function(s)
          protein kinase C binding protein binding zinc ion binding enzyme binding protein kinase binding protein binding, bridging receptor tyrosine kinase binding ubiquitin protein ligase binding ionotropic glutamate receptor binding signaling receptor activity SH2 domain binding identical protein binding ubiquitin binding macromolecular complex binding metal ion binding K63-linked polyubiquitin binding ubiquitin-dependent protein binding protein sequestering activity molecular sequestering activity molecular condensate scaffold activity protein homodimerization activity transcription cofactor activity
          Process(es)
          positive regulation of protein phosphorylation immune system process ubiquitin-dependent protein catabolic process autophagy apoptotic process macroautophagy positive regulation of macroautophagy cell differentiation regulation of I-kappaB kinase/NF-kappaB signaling protein heterooligomerization regulation of mitophagy negative regulation of transcription from RNA polymerase II promoter mitophagy mitophagy pexophagy temperature homeostasis response to ischemia transcription from RNA polymerase II promoter protein import into nucleus endosome organization protein localization positive regulation of autophagy regulation of mitochondrion organization endosomal transport protein catabolic process negative regulation of protein ubiquitination cellular response to stress toll-like receptor 4 signaling pathway negative regulation of toll-like receptor 4 signaling pathway intracellular signal transduction aggrephagy positive regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter regulation of Ras protein signal transduction regulation of protein complex stability brown fat cell proliferation protein K63-linked ubiquitination protein targeting to vacuole involved in autophagy energy homeostasis response to mitochondrial depolarisation negative regulation of ferroptosis membraneless organelle assembly positive regulation of long-term synaptic potentiation positive regulation of protein localization to plasma membrane protein localization to perinuclear region of cytoplasm mitophagy in response to mitochondrial depolarization regulation of nucleic acid-templated transcription
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline