    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ADNP Antibodies
          Invitrogen
          ADNP Polyclonal Antibody
          View all (13) ADNP antibodies
          Tech Support
          Cite ADNP Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          ADNP Polyclonal Antibody
          Product Details
          PA1-4223
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Synthetic peptides rcorresponding to residues M(1) F Q L P V N(7) C K(1096) L S S Q Q A(1102) of the N- and C-termini of human ADNP.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2222386
          Product Specific Information
          PA1-4223 detects the hADNP protein in human samples.
          PA1-4223 has been successfully used in Western blot procedures.
          The PA1-4223 immunogen is a synthetic peptides rcorresponding to residues M(1) F Q L P V N(7) C K(1096) L S S Q Q A(1102) of the N- and C-termini of human ADNP, separated by a cysteine.
          Target Information
          Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
          Bioinformatics
          Protein Aliases: Activity-dependent neuroprotective protein; activity-dependent neuroprotector; Activity-dependent neuroprotector homeobox protein; ADNP; ADNP homeobox 1; unnamed protein product
          View more View less
          Gene Aliases: ADNP; ADNP1; HVDAS; KIAA0784; MRD28
          View more View less
          UniProt ID: (Human) Q9H2P0
          View more View less
          Entrez Gene ID: (Human) 23394
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding chromatin binding copper ion binding protein binding beta-catenin binding zinc ion binding peptide binding metal ion binding beta-tubulin binding
          Process(es)
          regulation of transcription from RNA polymerase II promoter short-term memory response to carbohydrate regulation of gene expression negative regulation of gene expression positive regulation of neuron projection development cGMP-mediated signaling neuron differentiation nitric oxide homeostasis negative regulation of neuron apoptotic process estrous cycle positive regulation of axon extension negative regulation of synaptic transmission neuron apoptotic process positive regulation of synapse assembly positive regulation of canonical Wnt signaling pathway
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline