    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • hCG beta Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          CGB Polyclonal Antibody
          View all (80) hCG beta antibodies
          Tech Support
          Cite CGB Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CGB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CGB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CGB Antibody (PA5-109521) in WB
          Western blot analysis of CGB in MCF7, 293T, THP-1. Samples were incubated in polyclonal CGB antibody (Product # PA5-109521) using a dilution of 1:500, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10000. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CGB Antibody in Western Blot (WB)
          Product Details
          PA5-109521
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant protein of human CGB.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.19 mg/mL
          Amount
          19 µg
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Wet ice
          RRID
          AB_2854932
          Target Information
          hCG beta is the beta chain of hCG (Human chorionic gonadotropin), a 40 kDa glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. hCG is made up of alpha and beta subunits that are non-covalently associated. The alpha subunit is similar to those found on the pituitary hormones FSH, LH ans TSH. However, the beta subunit is different in FSH, LH and TSH and are involved in conferring biological specificity. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium. hCG is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). Diseases associated with Hcg dysfunction include gestational trophoblastic tumor and hydatidiform mole, recurrent 1.
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline