    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • GPR120 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          GPR120 (extracellular) Polyclonal Antibody
          View all (25) GPR120 antibodies
          Tech Support
          Cite GPR120 (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GPR120 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GPR120 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GPR120 (extracellular) Antibody (PA5-111778) in WB
          Western Blot analysis of GPR120 was performed in rat lung (lanes 1 and 3), rat tongue (lanes 2 and 4), human U-87 astrocytoma cells (lanes 5 and 7) and mouse MS1 endothelial cells (lanes 6 and 8) lysates. Lane 1,2,5,6: GPR120 (extracellular) Antibody (Product # PA5-111778) at a dilution of 1:200. Lane 3,4,7,8: GPR120 (extracellular) Antibody preincubated with the negative control antigen. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GPR120 (extracellular) Antibody in Western Blot (WB)
          GPR120 (extracellular) Antibody in Western Blot (WB)
          GPR120 (extracellular) Antibody in Flow Cytometry (Flow)
          GPR120 (extracellular) Polyclonal Antibody
          Product Details
          PA5-111778
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Flow Cytometry (Flow)
          2.5 µg/0.5x10^6 cells
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Peptide corresponding to amino acid residues 22-36 of mouse FFAR4 (Extracellular, N-terminus)
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          RRID
          AB_2857187
          Product Specific Information
          Reconstitute antibody with 50 µL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Peptide sequence is: (C)RTHFPFF SDVKGDHR
          Homology: Rat amino acid residues identical; human - 14/15 amino acid residues identical.
          Target Information
          An EST for GPR120 has been isolated from a human colon cancer library. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This "turn-off" activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline