  • Aplicaciones
    • Cultivo celular y transfección
    • Citometría de flujo
    • Investigación sobre el cáncer
    • Cromatografía
    • Secuenciación
    • PCR
    • Soluciones para laboratorio
    • Diagnóstico de alergias
    • Ver todas las aplicaciones y técnicas
  • Servicios
    • Servicios Personalizados
    • Servicios de Capacitación
    • Informática para laboratorios de ámbito empresarial
    • Servicios financieros y de arrendamiento
    • Servicios 360° de CDMO y CRO
    • Servicios de CDMO
    • Servicios de CRO
    • Ver todos los servicios
  • Ayuda y Soporte
    • Crear una nueva cuenta
    • Cómo hacer el pedido
    • Póngase en contacto con nosotros
    • Cambio de ubicación
    • Ver toda la ayuda y soporte técnico
      Cell Culture Plastics
  • A quiénes brindamos nuestros servicios
    • Industria biotecnológica
    • Sector biofarmacéutico
    • CDMO
    • Diagnósticos de laboratorio
    • Ciencias aplicadas e industriales
  • Ofertas especiales
  • Contáctenos
  • Orden Rápida
  • Documentos y certificados
Thermo Fisher Scientific Logo
Search
Buscar
Search button
          • Contáctenos
          • Orden Rápida
          • Iniciar sesión
            Iniciar sesión
            ¿No tiene una cuenta? Crear una cuenta
            • Cuenta
            • Pedidos
            • Productos y proyectos personalizados
          • Primary Antibodies ›
          • CHK2 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          Phospho-CHK2 (Ser19) Polyclonal Antibody
          View all (73) CHK2 antibodies
          Tech Support
          Cite Phospho-CHK2 (Ser19) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Phospho-CHK2 (Ser19) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-CHK2 (Ser19) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Phospho-CHK2 (Ser19) Antibody (PA5-118608) in ICC/IF
          Immunocytochemical analysis of Phospho-CHK2 (Ser19) in 293 cells using a Phospho-CHK2 (Ser19) polyclonal antibody (Product # PA5-118608). The samples were fixed with PFA and permeabilized in 0.1% Triton X-100,then blocked in 10% serum for 45 minutes at 25°C. The primary antibody was diluted at 1:200 and incubated with the sample for 1 hour at 37°C. An Alexa Fluor 594 conjugated goat anti-rabbit IgG (H+L) Ab... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-CHK2 (Ser19) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-CHK2 (Ser19) Antibody in Western Blot (WB)
          Phospho-CHK2 (Ser19) Polyclonal Antibody
          Product Details
          PA5-118608
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:3,000
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:500
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          A synthesized peptide derived from human Chk2 around the phosphorylation site of Ser19.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          RRID
          AB_2903108
          Target Information
          CHEK2 (CHK2) is a serine/threonine kinase and a component of the DNA damage checkpoint pathway. A mutation in CHK2 has been linked to cancer. CHK2 is activated by DNA damage and phosphorylates several modulators of cell cycle control including tumor suppressor proteins. In addition, Chk2 can phosphorylate BRCA1, allowing BRCA1 to restore survival after DNA damage. Chk2 is a putative tumor suppressor protein, with mutations in to the Chk2 gene linked to Li-Fraumeni syndrome, a familiar cancer usually associated with inherited mutations in TP53. Also, mutations in CHK2 are thought to confer a predisposition to sarcomas, breast cancer, and brain tumors.
          Pedidos Plus Icon Minus Icon
          • Estatus del pedido
          • Ayuda para pedidos
          • Orden Rápida
          • Supply Center
          • eProcurement
          Soporte Plus Icon Minus Icon
          • Ayuda y soporte
          • Entre en Contacto
          • Centros de asistencia técnica
          • Consultar documentos y certificados
          • Informar de un problema en la web
          Recursos Plus Icon Minus Icon
          • Centros de aprendizaje
          • Promociones
          • Eventos & Webinars
          • Medios Sociales
          Acerca de Thermo Fisher Plus Icon Minus Icon
          • Acerca de nosotros Acerca de nosotros
          • Empleo Empleo
          • Inversores Inversores
          • Noticias Noticias
          • Responsabilidad social Responsabilidad social
          • Marcas comerciales
          • Políticas y avisos
          Nuestro Portafolio Plus Icon Minus Icon
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline