    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SYNC Antibodies
          Invitrogen
          SYNC Polyclonal Antibody
          View all (13) SYNC antibodies
          Tech Support
          Cite SYNC Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          SYNC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SYNC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          SYNC Antibody (PA5-121568) in WB
          Western blot analysis of SYNC using SYNC Polyclonal Antibody (Product # PA5-121567) at a dilution of 2 µg/mL. Lane 1: Porcine Heart lysate; Lane 2: Rat Heart lysate; Lane 3: Mouse Heart lysate; Lane 4: Bovine Heart lysate. Secondary antibody used is HRP-Goat anti-Rabbit IgG polyclonal antibody at a dilution of 0.2 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SYNC Antibody in Western Blot (WB)
          SYNC Antibody in Western Blot (WB)
          SYNC Antibody in Western Blot (WB)
          SYNC Polyclonal Antibody
          Product Details
          PA5-121568
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.5-2 µg/mL
          -
          Immunohistochemistry (IHC)
          5-20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          5-20 µg/mL
          -
          Product Specifications
          Species Reactivity
          Bovine, Human, Mouse, Pig, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Ile169-Pro458 (Accession Q9H7C4), with N-terminal His and GST Tag
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915140
          Target Information
          This gene encodes a member of the intermediate filament family which contains an N-terminal head domain, followed by a central coiled-coil region and a short C-terminal tail. The protein is highly expressed in skeletal and cardiac muscle. The protein links the dystrophin associated protein complex (DAPC) to desmin filaments in muscle and may have a structural role in striated muscle. Multiple transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: intermediate filament protein syncoilin; synci; Syncoilin; Syncoilin intermediate filament 1; syncoilin, intermediate filament 1; Syncoilin-1; unnamed protein product
          View more View less
          Gene Aliases: 1110057H03Rik; SNIP4; SYNC; SYNC1; SYNCOILIN
          View more View less
          UniProt ID: (Human) Q9H7C4, (Mouse) Q9EPM5
          View more View less
          Entrez Gene ID: (Pig) 100620771, (Human) 81493, (Bovine) 618122, (Mouse) 68828, (Rat) 362606
          View more View less
          Function(s)
          protein binding structural molecule activity
          Process(es)
          intermediate filament-based process
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline