    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PXDN Antibodies
          Invitrogen
          PXDN Polyclonal Antibody
          View all (6) PXDN antibodies
          Tech Support
          Cite PXDN Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PXDN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PXDN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PXDN Antibody (PA5-121659) in WB
          Western blot analysis of PXDN in recombinant protein. Samples were incubated in polyclonal PXDN antibody (Product # PA5-121658). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PXDN Antibody in Western Blot (WB)
          PXDN Polyclonal Antibody
          Product Details
          PA5-121659
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-2 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Glu76-Gly260 (Accession Q92626), with N-terminal His Tag
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915231
          Target Information
          Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.
          Bioinformatics
          Protein Aliases: animal heme peroxidase; hsPxd01; KIAA0230; Melanoma-associated antigen MG50; p53-responsive gene 2 protein; Peroxidasin 1; Peroxidasin homolog; Vascular peroxidase 1
          View more View less
          Gene Aliases: ASGD7; COPOA; D2S448; D2S448E; hsPxd01; KIAA0230; MG50; PRG2; PXD01; PXDN; PXN; VPO; VPO1
          View more View less
          UniProt ID: (Human) Q92626
          View more View less
          Entrez Gene ID: (Human) 7837
          View more View less
          Function(s)
          peroxidase activity interleukin-1 receptor antagonist activity extracellular matrix structural constituent oxidoreductase activity oxidoreductase activity, acting on peroxide as acceptor heme binding laminin-1 binding metal ion binding lactoperoxidase activity
          Process(es)
          angiogenesis eye development negative regulation of cytokine-mediated signaling pathway immune response response to oxidative stress cell adhesion extracellular matrix organization collagen fibril organization hydrogen peroxide catabolic process system development protein homooligomerization protein homotrimerization basement membrane assembly basement membrane organization cellular oxidant detoxification
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline