    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PSAT1 Antibodies
          Invitrogen
          PSAT1 Polyclonal Antibody
          View all (22) PSAT1 antibodies
          Tech Support
          Cite PSAT1 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (7)
          PSAT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PSAT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          PSAT1 Antibody (PA5-121666) in ICC/IF
          Immunocytochemistry analysis of PSAT1 in human hela cells using PSAT1 Polyclonal Antibody (Product # PA5-119345). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PSAT1 Antibody in Immunocytochemistry (ICC/IF)
          PSAT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PSAT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PSAT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PSAT1 Antibody in Western Blot (WB)
          PSAT1 Antibody in Western Blot (WB)
          PSAT1 Antibody in Western Blot (WB)
          PSAT1 Polyclonal Antibody
          Product Details
          PA5-121666
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          5-20 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant protein Phosphoserine Aminotransferase 1. The antigen corresponds to amino acid range 1-312 of the target protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915238
          Target Information
          The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
          Bioinformatics
          Protein Aliases: Endometrial progesterone-induced protein; EPIP; Phosphohydroxythreonine aminotransferase; Phosphoserine aminotransferase; phosphoserine transaminase; PSAT; unnamed protein product
          View more View less
          Gene Aliases: D8Ertd814e; EPIP; NLS2; PSA; Psa1; PSAT; PSAT1; PSATD
          View more View less
          UniProt ID: (Human) Q9Y617, (Mouse) Q99K85
          View more View less
          Entrez Gene ID: (Human) 29968, (Pig) 100154160, (Rat) 293820, (Mouse) 107272
          View more View less
          Function(s)
          O-phospho-L-serine:2-oxoglutarate aminotransferase activity protein binding transaminase activity transferase activity pyridoxal phosphate binding identical protein binding catalytic activity transaminase
          Process(es)
          L-serine metabolic process L-serine biosynthetic process pyridoxine biosynthetic process cellular amino acid biosynthetic process metabolic process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline