          • Primary Antibodies ›
          • HLA-C Antibodies
          Invitrogen
          HLA-C Polyclonal Antibody
          View all (12) HLA-C antibodies
          Tech Support
          Cite HLA-C Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (13)
          HLA-C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HLA-C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 13
          HLA-C Antibody (PA5-121909) in ICC/IF
          Immunocytochemistry analysis of HLA-C in Human Hela cells using HLA-C Polyclonal Antibody (Product # PA5-119267) at a dilution of 10 µg/mL, followed by secondary antibody staining with 1 µg/mL FITC-Linked Caprine Anti-Rabbit IgG Polyclonal Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HLA-C Antibody in Immunocytochemistry (ICC/IF)
          HLA-C Antibody in Immunocytochemistry (ICC/IF)
          HLA-C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HLA-C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HLA-C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HLA-C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HLA-C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HLA-C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HLA-C Antibody in Western Blot (WB)
          HLA-C Antibody in Western Blot (WB)
          HLA-C Antibody in Western Blot (WB)
          HLA-C Antibody in Western Blot (WB)
          HLA-C Antibody in Western Blot (WB)
          HLA-C Polyclonal Antibody
          Product Details
          PA5-121909
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          5-20 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Pig
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant protein Major Histocompatibility Complex Class I C. The antigen corresponds to amino acid range 48-305 of the target protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915481
          Target Information
          HLA-C belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Over one hundred HLA-C alleles have been described.
          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
          Bioinformatics
          Protein Aliases: exons 2-8; HLA class I histocompatibility antigen, C alpha chain; HLA-C; HLA-C antigen; HLA-Cw; HLA-Cw1 protein; HLA-Cw2 protein; HLA-Cw3 protein; Human leukocyte antigen C; human leukocyte antigen-C alpha chain; major histocompatibility antigen HLA-C; major histocompatibility HLA-Cw2.2 heavy chain; MHC class I antigen heavy chain HLA-C; MHC HLA-C-alpha-2 chain precursor; unnamed protein product
          View more View less
          Gene Aliases: D6S204; HLA-C; HLA-JY3; HLAC; HLC-C; MHC; PSORS1
          View more View less
          UniProt ID: (Human) P10321
          View more View less
          Entrez Gene ID: (Human) 3107
          View more View less
          Function(s)
          receptor binding protein binding peptide antigen binding TAP binding receptor agonist activity
          Process(es)
          positive regulation of T cell mediated cytotoxicity adaptive immune response immune system process antigen processing and presentation of peptide antigen via MHC class I antigen processing and presentation of endogenous peptide antigen via MHC class Ib antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-independent immune response signal transduction antigen processing and presentation innate immune response positive regulation of natural killer cell mediated cytotoxicity
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline