  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • GSTA5 Antibodies
          Invitrogen
          GSTA5 Polyclonal Antibody
          View all (5) GSTA5 antibodies
          Tech Support
          Cite GSTA5 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GSTA5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GSTA5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GSTA5 Antibody (PA5-121940) in WB
          Western blot analysis of GSTA5 in Rat Liver lysate using GSTA5 Polyclonal Antibody (Product # PA5-119148) at a dilution of 2 µg/mL. Secondary antibody used is HRP-Goat anti-Rabbit IgG polyclonal antibody at a dilution of 0.2 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GSTA5 Antibody in Western Blot (WB)
          GSTA5 Antibody in Western Blot (WB)
          GSTA5 Antibody in Western Blot (WB)
          GSTA5 Polyclonal Antibody
          Product Details
          PA5-121940
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          5-20 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant protein Glutathione S Transferase Alpha 5. The antigen corresponds to amino acid range 1-221 of the target protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915512
          Target Information
          The glutathione S-transferases catalyze the conjugation of reduced glutathiones and a variety of electrophiles, including many known carcinogens and mutagens. The cytosolic GSTs belong to a large superfamily, with members located on different chromosomes.
          Bioinformatics
          Protein Aliases: GHS transferase; Glutathione S-transferase A5; Glutathione S-transferase A5-5; Glutathione S-transferase alpha-2; glutathione S-transferase Ya subunit; Glutathione S-transferase Ya-2; glutathione transferase YA subunit; GST 1b-1b; GST A2-2; GST class-alpha member 5; GST Ya2
          View more View less
          Gene Aliases: Gsta2; GSTA5
          View more View less
          UniProt ID: (Human) Q7RTV2, (Rat) P04903
          View more View less
          Entrez Gene ID: (Human) 221357, (Rat) 494499
          View more View less
          Function(s)
          glutathione transferase activity protein binding transferase activity transferase
          Process(es)
          glutathione metabolic process xenobiotic metabolic process metabolic process
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline