    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GDA Antibodies
          Invitrogen
          GDA Polyclonal Antibody
          View all (9) GDA antibodies
          Tech Support
          Cite GDA Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (9)
          GDA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GDA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          GDA Antibody (PA5-121962) in ICC/IF
          Immunocytochemistry analysis of GDA in Hela cells using GDA Polyclonal Antibody (Product # PA5-119167) at a dilution of 20 µg/mL, followed by secondary antibody staining with 2 µg/mL AF488-Linked Caprine Anti-Rabbit IgG Polyclonal Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GDA Antibody in Immunocytochemistry (ICC/IF)
          GDA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GDA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GDA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GDA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GDA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GDA Antibody in Western Blot (WB)
          GDA Antibody in Western Blot (WB)
          GDA Antibody in Western Blot (WB)
          GDA Polyclonal Antibody
          Product Details
          PA5-121962
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-3 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-30 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          5-30 µg/mL
          -
          Product Specifications
          Species Reactivity
          Bovine, Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant protein Guanine Deaminase. The antigen corresponds to amino acid range 1-454 of the target protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.91 mg/mL
          Amount
          18.2 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915534
          Target Information
          GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
          Bioinformatics
          Protein Aliases: cytoplasmic PSD-95 interactor; cytoplasmic PSD95 interactor; GAH; Guanase; guanine aminase; Guanine aminohydrolase; Guanine deaminase; NE-dlg binding protein; p51-nedasin; unnamed protein product
          View more View less
          Gene Aliases: AU015411; AW047581; CYPIN; GAH; GDA; GUANASE; KIAA1258; NEDASIN
          View more View less
          UniProt ID: (Human) Q9Y2T3, (Mouse) Q9R111
          View more View less
          Entrez Gene ID: (Bovine) 525937, (Human) 9615, (Mouse) 14544
          View more View less
          Function(s)
          zinc ion binding guanine deaminase activity hydrolase activity hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds metal ion binding protein binding deaminase
          Process(es)
          allantoin metabolic process nucleobase-containing compound metabolic process guanine catabolic process deoxyguanosine catabolic process nervous system development cellular amide catabolic process GMP catabolic process dGMP catabolic process guanine metabolic process positive regulation of microtubule polymerization
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline