    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • PLA2G4A Antibodies
          Invitrogen
          Phospho-PLA2G4A (Ser505) Polyclonal Antibody
          Advanced Verification
          View all (17) PLA2G4A antibodies
          Tech Support
          Cite Phospho-PLA2G4A (Ser505) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Advanced Verification (1)
          Phospho-PLA2G4A (Ser505) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PLA2G4A (Ser505) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Phospho-PLA2G4A (Ser505) Antibody (PA5-17790) in WB
          Western blot was performed using Anti-PLA2G4A Polyclonal Antibody (Product # PA5-17790) and a ~85 kDa band was observed across the panel tested except IMR-32 and Reh. Whole cell extracts (30 µg lysate) of HeLa (Lane 1), A549 (Lane 2), NIH/3T3 (Lane 3), IMR-32 (Lane 4) and Reh (Lane 5) were electrophoresed using NuPAGE® 4-12 % Bis-Tris gel (Product # NP0322BOX). Resolved proteins were then transferred onto a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-PLA2G4A (Ser505) Antibody in Western Blot (WB)
          Phospho-PLA2G4A (Ser505) Antibody
          Phospho-PLA2G4A (Ser505) Polyclonal Antibody
          Product Details
          PA5-17790
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunoprecipitation (IP)
          1:50
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Non-human primate, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Synthetic phosphopeptide corresponding to residues surrounding pSer505 of human cPLA2
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          250 µg/mL
          Amount
          25 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol
          Contains
          no preservative
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_10985549
          Product Specific Information
          It is not recommended to aliquot this antibody.
          This antibody is not cross-reactive with the secreted PLA2 (sPLA2) or cytosolic Ca2+ independent PLA2 (iPLA2).
          Target Information
          This gene encodes a member of the cytosolic phospholipase A2 group IV family. The enzyme catalyzes the hydrolysis of membrane phospholipids to release arachidonic acid which is subsequently metabolized into eicosanoids. Eicosanoids, including prostaglandins and leukotrienes, are lipid-based cellular hormones that regulate hemodynamics, inflammatory responses, and other intracellular pathways. The hydrolysis reaction also produces lysophospholipids that are converted into platelet-activating factor. The enzyme is activated by increased intracellular Ca(2+) levels and phosphorylation, resulting in its translocation from the cytosol and nucleus to perinuclear membrane vesicles.
          Bioinformatics
          Protein Aliases: calcium-dependent phospholipid-binding protein; cPLA2; cytosolic group IVA; Cytosolic phospholipase A2; group IVA; lysophospholipase; MGC126350; phosphatidylcholine 2-acylhydrolase; phospholipase A2; phospholipase A2 cytosolic; Phospholipase A2 group IVA; phospholipase A2, group IVA (cytosolic, calcium-dependent); unnamed protein product
          View more View less
          Gene Aliases: CPLA2; cPLA2-alpha; cPLA2alpha; GURDP; Pla2c; PLA2G4; PLA2G4A
          View more View less
          UniProt ID: (Human) P47712, (Mouse) P47713
          View more View less
          Entrez Gene ID: (Human) 5321, (Mouse) 18783, (Rat) 24653
          View more View less
          Function(s)
          phospholipase activity lysophospholipase activity phospholipase A2 activity calcium ion binding calcium-dependent phospholipid binding lipid binding O-acyltransferase activity phosphatidylinositol-5-phosphate binding hydrolase activity phosphatidylinositol-3-phosphate binding metal ion binding calcium-dependent phospholipase A2 activity calcium-independent phospholipase A2 activity carboxylic ester hydrolase activity phosphatidylinositol-4-phosphate binding ceramide 1-phosphate binding histone acetyltransferase binding phospholipase
          Process(es)
          prostaglandin biosynthetic process positive regulation of T-helper 1 type immune response glycerol metabolic process lipid metabolic process fatty acid metabolic process fatty acid biosynthetic process monoacylglycerol biosynthetic process glycerophospholipid metabolic process platelet activating factor biosynthetic process icosanoid metabolic process prostaglandin metabolic process phospholipid catabolic process positive regulation of platelet activation lipid catabolic process arachidonic acid metabolic process leukotriene biosynthetic process positive regulation of prostaglandin secretion phosphatidylglycerol catabolic process phosphatidylcholine catabolic process phosphatidylcholine acyl-chain remodeling regulation of cell proliferation positive regulation of macrophage activation icosanoid biosynthetic process glycerophospholipid catabolic process arachidonic acid secretion establishment of localization in cell cellular response to antibiotic ovulation from ovarian follicle luteolysis aging positive regulation of cell proliferation response to heat response to hormone response to organic substance response to lithium ion response to organonitrogen compound positive regulation of bone mineralization positive regulation of vesicle fusion positive regulation of prostaglandin biosynthetic process positive regulation of fever generation response to lipopolysaccharide response to vitamin D response to hydrogen peroxide positive regulation of apoptotic process surfactant homeostasis decidualization positive regulation of inflammatory response response to glucocorticoid response to calcium ion response to methylmercury
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline