    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DACT1 Antibodies
          Invitrogen
          DACT1 Polyclonal Antibody
          1 Reference
          View all (10) DACT1 antibodies
          Tech Support
          Cite DACT1 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          DACT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          DACT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          DACT1 Antibody (PA5-23216) in IHC (P)
          Immunohistochemical analysis of DACT1 in formalin-fixed paraffin-embedded human brain. Samples were incubated in DACT1 polyclonal antibody (Product # PA5-23216) using a dilution of 10 µg/mL. Staining of formalin-fixed tissues is enhanced by boiling tissue sections in 10 mM sodium citrate buffer, pH 6.0 for 10-20 min followed by cooling at RT for 20 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DACT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DACT1 Antibody in Western Blot (WB)
          DACT1 Polyclonal Antibody
          Product Details
          PA5-23216
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.5-1 µg/mL
          View 1 publication 1 publication
          Immunohistochemistry (Paraffin) (IHC (P))
          10 µg/mL
          -
          Product Specifications
          Species Reactivity
          Chicken, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
          Published species
          Zebrafish
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          A portion of amino acids 725-775 of human Dapper homolog 1.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2540741
          Target Information
          Dapper homolog 1, an ortholog of Xenopus Dpr, antagonizes Wnt signaling by promoting Dvl degradation via a lysosome inhibitor-sensitive and proteasome inhibitor-insensitive mechanism. Dapper homolog 1 acts as an interacting protein for Dvl, and the interaction is formed between the DEP (Dishevelled, Egl-10, and pleckstrin) domain of Dvl and the central and C-terminal regions of Dapper homolog 1. The gene for human Dapper homolog 1 is localized on chromosomal region 14q23.1. The protein impedes the degradation of CTNNB1/beta-catenin, thereby enhancing the transcriptional activation of target genes of the Wnt signaling pathway. The protein is also required for normal notochord formation and is downregulated in hepatocellular carcinomas (HCC). It is expressed in human liver and various other tissues.
          Bioinformatics
          Protein Aliases: dapper 1; Dapper antagonist of catenin 1; Dapper homolog 1; dapper homolog 1, antagonist of beta-catenin; dapper, antagonist of beta-catenin; dapper, antagonist of beta-catenin, homolog 1; dapper1b; dishevelled-binding antagonist of beta-catenin 1; dishevelled-interacting protein; Frodo; frodo 1; Frodo homolog; functional regulator of dsh in ontogenesis; hDPR1; Hepatocellular carcinoma novel gene 3 protein; heptacellular carcinoma novel gene 3; HNG3; MDpr1; signaling protein; thymus expressed gene 3; Thymus-expressed novel gene 3 protein; unnamed protein product
          View more View less
          Gene Aliases: 4921528D17Rik; AI115603; DACT1; dact1-a; dact1-b; DAPPER; DAPPER1; dpr; DPR1; Frd1; FRODO; Frodo1; HDPR1; HNG3; MDpr1; MTNG3; RGD1564008; TBS2; THYEX3; XDpr; XDpr1b; XELAEV_18041122mg
          View more View less
          UniProt ID: (Human) Q9NYF0, (Mouse) Q8R4A3
          View more View less
          Entrez Gene ID: (Human) 51339, (Chicken) 723789, (Rat) 500666, (Mouse) 59036, (Zebrafish) 405799, (Xenopus) 398315
          View more View less
          Function(s)
          protein kinase C binding protein binding beta-catenin binding histone deacetylase binding protein kinase A binding RNA polymerase II sequence-specific DNA binding transcription factor binding delta-catenin binding transcription regulator inhibitor activity
          Process(es)
          multicellular organism development Wnt signaling pathway negative regulation of JNK cascade negative regulation of canonical Wnt signaling pathway regulation of Wnt signaling pathway, planar cell polarity pathway negative regulation of transcription from RNA polymerase II promoter nervous system development neural tube development positive regulation of Wnt signaling pathway negative regulation of Wnt signaling pathway regulation of protein stability positive regulation of protein catabolic process embryonic hindgut morphogenesis regulation of canonical Wnt signaling pathway positive regulation of canonical Wnt signaling pathway regulation of nodal signaling pathway negative regulation of beta-catenin-TCF complex assembly negative regulation of G1/S transition of mitotic cell cycle gastrulation with mouth forming second regulation of catenin import into nucleus positive regulation of catenin import into nucleus positive regulation of fat cell differentiation embryonic morphogenesis dendrite morphogenesis synapse organization canonical Wnt signaling pathway
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline